This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00444645_g1
See other USO1 GE Assays ›
Gene Symbol:
Uso1
Gene Name:
USO1 vesicle docking factor
Gene Aliases:
115kDa, TAP, Vdp
Chromosome Location:
Chr.5: 92137909 - 92202800 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00444645_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Uso1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_019490.1 NP_062363.1 6 - 7 661 90
GenBank mRNA AF096868.1 - 5 - 6 449 90
AK144638.1 - 6 - 7 652 90
AK156070.1 - 6 - 7 654 90
AK166587.1 - 7 - 8 621 90
AK172425.1 - 6 - 7 652 90
BC016069.1 - 6 - 7 661 4480992 90

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Uso1

Panther Classification:

Molecular Function -

membrane traffic protein

Gene Ontology Categories:

Function(s) Process(es)

transport
intracellular protein transport
ER to Golgi vesicle-mediated transport
protein transport
vesicle-mediated transport
transcytosis
Golgi vesicle transport
Golgi vesicle docking
vesicle fusion with Golgi apparatus
membrane fusion
protein binding
protein transporter activity
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-4krp5:80/100.66.77.213:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0