This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00445227_m1
See other FABP7 GE Assays ›
Gene Symbol:
Fabp7
Gene Name:
fatty acid binding protein 7, brain
Gene Aliases:
B-FABP, BFABP, Blbp, MRG
Chromosome Location:
Chr.10: 57784923 - 57788450 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00445227_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Fabp7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_021272.3 NP_067247.1 3 - 4 433 86
GenBank mRNA AJ870970.1 - 3 - 4 433 86
AK002955.1 - 3 - 4 434 86
AK021271.1 - 3 - 4 438 86
BC055280.1 - 3 - 4 416 5710017 86
BC057090.1 - 3 - 4 419 5700428 86
CT010401.1 - 3 - 4 347 86
S69799.1 - 3 - 4 416 86

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000000508.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Fabp7

Gene Ontology Categories:

Function(s) Process(es)

startle response
transport
cell proliferation in forebrain
neurogenesis
epithelial cell proliferation
prepulse inhibition
transporter activity
fatty acid binding
lipid binding

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P290074 S69799.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P290074 AK002955.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P290074 AK021271.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P290074 BC055280.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P290074 NM_021272.3 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P290074 S69799.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P290074 AK002955.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P290074 AK021271.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P290074 BC055280.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P290074 NM_021272.3 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 768352 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P166196 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P290074 S69799.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P290074 AK002955.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P290074 AK021271.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P290074 BC055280.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P290074 NM_021272.3 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P290074 S69799.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P290074 AK002955.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P290074 AK021271.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P290074 BC055280.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P290074 NM_021272.3 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A ERCC-00003_63 - -
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P290074 S69799.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P290074 AK002955.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P290074 AK021271.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P290074 BC055280.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P290074 NM_021272.3 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P290074 S69799.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P290074 AK002955.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P290074 AK021271.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P290074 BC055280.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P290074 NM_021272.3 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27162236 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27162210 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27162196 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27162206 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27162209 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27162193 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27162231 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27162238 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27162227 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27162195 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27162237 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27162229 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27162240 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27162181 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27162213 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27162216 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27162234 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27162186 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27162187 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27162184 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27162211 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27162188 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27162224 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27162208 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27162232 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27162218 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27162220 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27162191 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27162185 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27162182 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27162235 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27162222 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P166196 - -
Whole Mouse Genome G4122A A_51_P290074 S69799.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P290074 AK002955.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P290074 AK021271.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P290074 BC055280.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P290074 NM_021272.3 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P290074 S69799.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P290074 AK002955.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P290074 AK021271.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P290074 BC055280.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P290074 NM_021272.3 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130037 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P166196 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08612660 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08612652 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08612642 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08585036 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08612648 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08612637 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162227 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162229 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162207 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162265 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162282 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162151 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162160 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162218 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162238 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162237 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162312 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162180 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162249 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162163 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162275 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162291 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162166 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162169 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162296 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162220 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162301 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162279 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162203 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162308 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162284 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162187 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162267 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162213 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162299 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162240 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162161 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162210 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162276 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162191 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162268 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162262 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162242 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162171 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162248 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162212 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162281 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162182 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162157 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162164 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162176 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162189 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162277 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162236 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162288 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162309 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162184 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162195 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162175 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162222 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162158 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162293 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162272 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162188 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162270 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162234 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162266 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162172 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162178 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162193 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162231 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162185 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162155 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162304 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162246 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162224 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162174 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162286 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162295 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162243 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27162216 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27162193 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27162180 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27162203 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27162238 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27162243 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27162236 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27162220 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08612651 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27162218 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27162167 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27162246 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27162231 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27162195 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27162206 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08847856 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27162227 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27162182 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27162188 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27162173 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27162240 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27162229 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08585034 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27162171 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27162234 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27162224 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08612661 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08612660 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27162212 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27162175 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27162169 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27162174 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27162191 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27162242 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27162210 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27162172 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27162178 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27162248 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27162196 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08206172 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27162185 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27162187 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27162235 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08612652 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27162208 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27162237 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08332068 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10363229 - -
Affymetrix Mouse Gene 1.0 ST Array 10363226 - -
Affymetrix Mouse Gene 1.0 ST Array 10363230 - -
Affymetrix Mouse Gene 1.0 ST Array 10363227 - -
Affymetrix Mouse Gene 1.0 ST Array 10363225 - -
Affymetrix Mouse Gene 1.0 ST Array 10363228 - -
Affymetrix HT MG-430 PM Array Plate 1450779_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10363224 - -
Mouse Genome 430 2.0 Array 1450779_PM_at - -
Mouse Genome 430 2.0 Array 1450779_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 12140_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 12140_at - -
Affymetrix Mouse Clariom S Array PSR1000003314.mm.1 NM_021272.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000003314.mm.1 NM_021272.3 Refseq
Affymetrix Mouse Clariom S Array PSR1000003314.mm.1 TC1000000508.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1000003315.mm.1 NM_021272.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000003315.mm.1 NM_021272.3 Refseq
Affymetrix Mouse Clariom S Array PSR1000003315.mm.1 TC1000000508.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1000003317.mm.1 NM_021272.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000003317.mm.1 NM_021272.3 Refseq
Affymetrix Mouse Clariom S Array PSR1000003317.mm.1 TC1000000508.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1000003313.mm.1 NM_021272.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000003313.mm.1 NM_021272.3 Refseq
Affymetrix Mouse Clariom S Array PSR1000003313.mm.1 TC1000000508.mm.1 MTA TC
Mouse Expression Set 430 1450779_at - -
Mouse Expression Set 430 1450779_PM_at - -
Murine Genome U74v2 Set 98967_at - -
Murine Genome U74v2 Set 98967_PM_at - -
Murine Genome U74v2 Set 169780_PM_i_at - -
Murine Genome U74v2 Set 166867_f_at - -
Murine Genome U74v2 Set 169780_i_at - -
Murine Genome U74v2 Set 166867_PM_f_at - -
Mouse Genome 430A 2.0 Array 1450779_at - -
Mouse Genome 430A 2.0 Array 1450779_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1450779_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1450779_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 775121 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 4590142 - -
Illumina mouse-6 v1.0 expression beadchip 4590142 - -
Illumina mouseRef-8 v1.0 expression beadchip 0004590142 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_1245241 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1245241 - -
mouse-6 v1.1 (Illumina) scl38907.5.1_34-S - -
Illumina Sentrix mouse-6.v1 bead array 4590142 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl38907.5.1_34-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 4590142 - -
MouseRef-8 v2 Expression BeadChip ILMN_2738330 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2738330 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0