Assay Details
| Gene Symbol: Rps6kb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021485.2 | NP_067460.1 | 13 - 14 | 1155 | 60 | |
| GenBank mRNA | AJ007938.1 | - | 13 - 14 | 1155 | 60 | |
| AK014412.1 | - | 13 - 14 | 1234 | 60 | ||
| AK090093.1 | - | 1210 | 60 | |||
| AK156361.1 | - | 13 - 14 | 1209 | 60 | ||
| BC005694.1 | - | 5 - 6 | 486 | 60 | ||
| BC051631.1 | - | 13 - 14 | 1461 | 60 | ||
| BC066842.1 | - | 12 - 13 | 1572 | 30468251 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rps6kb2
Genomic Map