This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00445525_g1
See other STARD3 GE Assays ›
Gene Symbol:
Stard3
Gene Name:
START domain containing 3
Gene Aliases:
Mln64, es64
Chromosome Location:
Chr.11: 98358367 - 98381112 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00445525_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Stard3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_021547.3 NP_067522.1 6 - 7 741 63
GenBank mRNA AK041538.1 - 6 - 7 697 63
AK150166.1 - 6 - 7 685 63
AK150254.1 - 6 - 7 685 63
AK155124.1 - 6 - 7 680 63
AK160480.1 - 6 - 7 661 63
AK205267.1 - 2 - 3 191 63
BC003313.1 - 6 - 7 658 3155179 63
BC058727.1 - 7 - 8 828 63
X82457.1 - 6 - 7 725 63

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100001565.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Stard3

Gene Ontology Categories:

Function(s) Process(es)

steroid biosynthetic process
progesterone biosynthetic process
transport
lipid transport
molecular_function
lipid binding
cholesterol binding
cholesterol transporter activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0