This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00445527_g1
See other STARD3 GE Assays ›
Gene Symbol:
Stard3
Gene Name:
START domain containing 3
Gene Aliases:
Mln64, es64
Chromosome Location:
Chr.11: 98358367 - 98381112 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00445527_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Stard3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_021547.3 NP_067522.1 8 - 9 896 61
GenBank mRNA AK041538.1 - 8 - 9 852 61
AK150166.1 - 8 - 9 840 61
AK150254.1 - 8 - 9 840 61
AK155124.1 - 8 - 9 835 61
AK160480.1 - 8 - 9 816 61
BC003313.1 - 8 - 9 813 3155179 61
BC058727.1 - 9 - 10 983 61
X82457.1 - 8 - 9 880 61

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100001565.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Stard3

Gene Ontology Categories:

Function(s) Process(es)

steroid biosynthetic process
progesterone biosynthetic process
transport
lipid transport
molecular_function
lipid binding
cholesterol binding
cholesterol transporter activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0