This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00445532_m1
See other STARD3 GE Assays ›
Gene Symbol:
Stard3
Gene Name:
START domain containing 3
Gene Aliases:
Mln64, es64
Chromosome Location:
Chr.11: 98358367 - 98381112 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00445532_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Stard3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_021547.3 NP_067522.1 13 - 14 1331 86
GenBank mRNA AK041538.1 - 13 - 14 1287 86
AK155124.1 - 13 - 14 1270 86
BC003313.1 - 13 - 14 1248 3155179 86
BC058727.1 - 14 - 15 1418 86
X82457.1 - 13 - 14 1315 86

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100001565.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Stard3

Gene Ontology Categories:

Function(s) Process(es)

steroid biosynthetic process
progesterone biosynthetic process
transport
lipid transport
molecular_function
lipid binding
cholesterol binding
cholesterol transporter activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0