Assay Details
| Gene Symbol: Gpr35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104529.1 | NP_001097999.1 | 1 - 2 | 168 | 71 | |
| GenBank mRNA | AF200349.1 | - | 1 - 2 | 89 | 71 | |
| AK036503.1 | - | 1 - 2 | 166 | 71 | ||
| AK154203.1 | - | 1 - 2 | 91 | 71 | ||
| BY233205.1 | - | 1 - 2 | 169 | 71 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF200349.1,AK036503.1,NM_001104529.1 transcripts.
Set Membership: |
|

Gpr35
Genomic Map