This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00445621_m1
See other FGF23 GE Assays ›
Gene Symbol:
Fgf23
Gene Name:
fibroblast growth factor 23
Gene Aliases:
-
Chromosome Location:
Chr.6: 127072902 - 127082296 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00445621_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Fgf23
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_022657.4 NP_073148.1 1 - 2 335 98
GenBank mRNA AB037889.1 - 1 - 2 207 98
AF263536.1 - 1 - 2 263 98
AK155687.1 - 1 - 2 335 98
BC120605.1 - 1 - 2 230 40129528 98

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600001476.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Fgf23

Panther Classification:

Molecular Function -

growth factor intercellular signal molecule

Gene Ontology Categories:

Function(s) Process(es)

fibroblast growth factor receptor signaling pathway
regulation of phosphate transport
positive regulation of vitamin D 24-hydroxylase activity
cell differentiation
regulation of bone mineralization
negative regulation of bone mineralization
cellular phosphate ion homeostasis
vitamin D catabolic process
negative regulation of osteoblast differentiation
positive regulation of transcription, DNA-templated
negative regulation of hormone secretion
phosphate ion homeostasis
positive regulation of ERK1 and ERK2 cascade
positive regulation of MAPKKK cascade by fibroblast growth factor receptor signaling pathway
fibroblast growth factor receptor binding
type 1 fibroblast growth factor receptor binding
protein binding
growth factor activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24830596 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24830595 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24830594 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24830598 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24830597 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2169514 AF263536.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2169514 AK155687.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2169514 NM_022657.3 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1976655 AK155687.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1976655 NM_022657.3 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 7265 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 937392 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 465990 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 952716 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 174907 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P179965 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2169514 AF263536.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2169514 AK155687.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2169514 NM_022657.3 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1976655 AK155687.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1976655 NM_022657.3 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P192523 BC120605.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P192523 AB037889.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P192521 AF263536.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P192521 AK155687.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P192521 BC120605.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P192521 NM_022657.3 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P192521 AB037889.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24830577 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24830566 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24830573 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24830535 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24830567 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24830584 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24830553 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24830557 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24830571 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24830572 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24830538 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24830537 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24830532 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24830536 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24830540 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24830560 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24830568 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24830583 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24830550 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24830562 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24830570 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24830542 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24830552 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24830576 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24830544 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24830548 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24830547 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24830579 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24830555 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24830545 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24830564 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24830554 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24830563 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24830534 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06062295 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P179965 - -
Whole Mouse Genome G4122A A_51_P192521 AF263536.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P192521 AK155687.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P192521 BC120605.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P192521 NM_022657.3 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P192521 AB037889.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P192523 BC120605.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P192523 AB037889.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P121811 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P179965 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05268315 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05268320 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05268310 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05268325 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05268312 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05268318 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05268319 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24830598 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24830596 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24830597 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24830594 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24830595 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830608 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830579 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830538 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830518 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830568 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830611 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830528 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830535 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830542 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830617 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830563 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830566 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830610 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830570 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830520 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830572 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830603 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830546 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830605 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830515 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830606 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830554 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830599 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830555 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830510 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830577 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830564 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830522 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830602 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830592 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830562 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830560 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830598 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830589 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830544 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830591 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830545 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830536 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830614 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830509 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830616 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830600 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830531 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830557 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830553 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830540 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830534 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830550 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830524 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830583 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830537 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830594 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830612 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830588 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830593 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24830586 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05797764 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24830593 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24830530 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24830606 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24830588 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24830574 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24830584 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24830528 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24830535 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24830594 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24830572 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24830526 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24830555 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24830566 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05663608 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24830557 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24830513 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24830536 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05489041 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24830610 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05806028 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24830512 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05739714 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24830568 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24830583 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24830542 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24830520 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24830538 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24830524 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24830603 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24830560 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05268315 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24830612 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24830614 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05355495 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24830611 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24830617 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24830563 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24830590 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24830616 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24830608 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24830570 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05268318 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24830537 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24830577 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05421335 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05489038 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24830515 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05314020 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24830540 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24830534 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24830518 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24830548 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24830605 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24830579 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24830586 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24830531 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10542029 - -
Affymetrix Mouse Gene 1.0 ST Array 10542032 - -
Affymetrix Mouse Gene 1.0 ST Array 10542030 - -
Affymetrix Mouse Gene 1.0 ST Array 10542031 - -
Affymetrix Mouse Gene 1.0 ST Array 10542033 - -
Affymetrix HT MG-430 PM Array Plate 1422176_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10542028 - -
Mouse Genome 430 2.0 Array 1422176_at - -
Mouse Genome 430 2.0 Array 1422176_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 64654_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 64654_at - -
Affymetrix Mouse Clariom S Array PSR0600012173.mm.1 NM_022657.4 Refseq
Affymetrix Mouse Clariom S Array PSR0600012173.mm.1 TC0600001476.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600012172.mm.1 NM_022657.4 Refseq
Affymetrix Mouse Clariom S Array PSR0600012172.mm.1 TC0600001476.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600012174.mm.1 NM_022657.4 Refseq
Affymetrix Mouse Clariom S Array PSR0600012174.mm.1 TC0600001476.mm.1 MTA TC
Mouse Expression Set 430 1422176_at - -
Mouse Expression Set 430 1422176_PM_at - -
Mouse Genome 430A 2.0 Array 1422176_PM_at - -
Mouse Genome 430A 2.0 Array 1422176_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 802412 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 510746 - -
Illumina mouse-6 v1.0 expression beadchip 510746 - -
Illumina mouseRef-8 v1.0 expression beadchip 0000510746 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2687816 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2687816 - -
mouse-6 v1.1 (Illumina) scl29484.2.1_23-S - -
Illumina Sentrix mouse-6.v1 bead array 510746 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl29484.2.1_23-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 510746 - -
MouseRef-8 v2 Expression BeadChip ILMN_2687816 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2687816 - -
Illumina Sentrix Mouse-6 Expression BeadChip (3) scl29484.2.1_23-S - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline