This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00445970_m1
See other ABCG8 GE Assays ›
Gene Symbol:
Abcg8
Gene Name:
ATP-binding cassette, sub-family G (WHITE), member 8
Gene Aliases:
1300003C16Rik, AI114946, sterolin-2
Chromosome Location:
Chr.17: 84676302 - 84700333 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00445970_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information
  • Citations (1)

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Abcg8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026180.3 NP_080456.1 1 - 2 172 70
GenBank mRNA AF324495.1 - 1 - 2 162 70
AK004871.1 - 1 - 2 129 70

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1700001265.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mus musculus Mouse ABC Transporters 1700 Inventoried Mouse_ABC_Transporters Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Abcg8

Panther Classification:

Molecular Function -

ATP-binding cassette (ABC) transporter primary active transporter transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
excretion
negative regulation of intestinal phytosterol absorption
phospholipid transport
sterol transport
intestinal cholesterol absorption
cholesterol efflux
cholesterol homeostasis
negative regulation of intestinal cholesterol absorption
sterol homeostasis
ATP binding
sterol transporter activity
ATPase activity
cholesterol transporter activity
protein heterodimerization activity

MicroArray Probes:

ABI

Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 570984 - -

Back To Top
 

Citations (1)

Fibroblast growth factor 21 corrects obesity in mice.
Authors
Coskun T, Bina HA, Schneider MA, Dunbar JD, Hu CC, Chen Y, Moller DE, Kharitonenkov A
Journal
Endocrinology - Page No 6018 (Mon Dec 01 00:00:00 EST 2008)
Volume (Year)
149 (2008)
Product Usage
-

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-9hc65:80/100.66.78.230:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0