This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00445973_g1
See other ABCG8 GE Assays ›
Gene Symbol:
Abcg8
Gene Name:
ATP-binding cassette, sub-family G (WHITE), member 8
Gene Aliases:
1300003C16Rik, AI114946, sterolin-2
Chromosome Location:
Chr.17: 84676302 - 84700333 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00445973_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Abcg8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001286005.1 NP_001272934.1 3 - 4 423 101
NM_026180.3 NP_080456.1 4 - 5 675 101
NR_104382.1 4 - 5 672 101
GenBank mRNA AF324495.1 - 4 - 5 665 101
AK004871.1 - 4 - 5 632 101
AK050938.1 - 3 - 4 425 101
AY196215.1 - 4 - 5 662 101
AY196216.1 - 4 - 5 662 101
BC138484.1 - 4 - 5 639 8861504 101
BC145293.1 - 4 - 5 639 9053847 101

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1700001265.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Abcg8

Panther Classification:

Molecular Function -

ATP-binding cassette (ABC) transporter primary active transporter transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
excretion
negative regulation of intestinal phytosterol absorption
phospholipid transport
sterol transport
intestinal cholesterol absorption
cholesterol efflux
cholesterol homeostasis
negative regulation of intestinal cholesterol absorption
sterol homeostasis
ATP binding
sterol transporter activity
ATPase activity
cholesterol transporter activity
protein heterodimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0