This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00446185_m1
See other IL1RN GE Assays ›
Gene Symbol:
Il1rn
Gene Name:
interleukin 1 receptor antagonist
Gene Aliases:
F630041P17Rik, IL-1ra
Chromosome Location:
Chr.2: 24336860 - 24351491 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00446185_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information
  • Citations (1)

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Il1rn
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_031167.5 NP_112444.1 1 - 2 132 75
GenBank mRNA AF001795.1 - 1 - 2 133 75
AK076296.1 - 1 - 2 133 75
AK170378.1 - 1 - 2 138 75
BC042532.1 - 1 - 2 4486098 75

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200000382.mm.1

Additional Information:

This assay is in the 5'UTR region of the AF001795.1,AK076296.1,AK170378.1,BC042532.1,NM_031167.5 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Il1rn

Panther Classification:

Molecular Function -

interleukin superfamily

Gene Ontology Categories:

Function(s) Process(es)

fever generation
negative regulation of cytokine-mediated signaling pathway
lipid metabolic process
acute-phase response
memory
negative regulation of glutamate secretion
cytokine-mediated signaling pathway
sensory perception of pain
insulin secretion
negative regulation of cell migration
negative regulation of heterotypic cell-cell adhesion
negative regulation of apoptotic process
positive regulation of JUN kinase activity
negative regulation of membrane potential
response to glucocorticoid
extracellular negative regulation of signal transduction
negative regulation of receptor activity
negative regulation of interleukin-1-mediated signaling pathway
cytokine activity
interleukin-1 receptor binding
interleukin-1, Type I receptor binding
interleukin-1, Type II receptor binding
interleukin-1 receptor antagonist activity
interleukin-1 Type I receptor antagonist activity
interleukin-1 Type II receptor antagonist activity

Back To Top
 

Citations (1)

Accelerated recovery from acute hypoxia in obese mice is due to obesity-associated up-regulation of interleukin-1 receptor antagonist.
Authors
Sherry CL, Kim SS, Freund GG
Journal
Endocrinology - Page No 2660 (Mon Jun 01 00:00:00 EDT 2009)
Volume (Year)
150 (2009)
Product Usage
-

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0