This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00446186_m1
See other IL1RN GE Assays ›
Gene Symbol:
Il1rn
Gene Name:
interleukin 1 receptor antagonist
Gene Aliases:
F630041P17Rik, IL-1ra
Chromosome Location:
Chr.2: 24336860 - 24351491 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00446186_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Il1rn
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001039701.3 NP_001034790.1 1 - 2 198 80
NM_001159562.1 NP_001153034.1 2 - 3 127 80
NM_031167.5 NP_112444.1 2 - 3 182 80
GenBank mRNA AF001795.1 - 2 - 3 183 80
AK076296.1 - 2 - 3 183 80
AK089222.1 - 1 - 2 153 80
AK154601.1 - 1 - 2 153 80
AK155263.1 - 1 - 2 153 80
AK155641.1 - 1 - 2 199 80
AK155756.1 - 1 - 2 153 80
AK155798.1 - 1 - 2 153 80
AK155846.1 - 1 - 2 153 80
AK155888.1 - 1 - 2 195 80
AK155908.1 - 1 - 2 153 80
AK156034.1 - 1 - 2 153 80
AK157157.1 - 1 - 2 191 80
AK170378.1 - 2 - 3 188 80
AK171037.1 - 2 - 3 131 80
AK171385.1 - 1 - 2 186 80
BC042532.1 - 2 - 3 159 4486098 80
BY206809.1 - 2 - 3 128 80
M57525.1 - 1 - 2 131 80
M64404.1 - 1 - 2 213 80
M74294.1 - 1 - 2 164 80
S64082.1 - 1 - 2 50 80

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200000382.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Il1rn

Panther Classification:

Molecular Function -

interleukin superfamily

Gene Ontology Categories:

Function(s) Process(es)

fever generation
negative regulation of cytokine-mediated signaling pathway
lipid metabolic process
acute-phase response
memory
negative regulation of glutamate secretion
cytokine-mediated signaling pathway
sensory perception of pain
insulin secretion
negative regulation of cell migration
negative regulation of heterotypic cell-cell adhesion
negative regulation of apoptotic process
positive regulation of JUN kinase activity
negative regulation of membrane potential
response to glucocorticoid
extracellular negative regulation of signal transduction
negative regulation of receptor activity
negative regulation of interleukin-1-mediated signaling pathway
cytokine activity
interleukin-1 receptor binding
interleukin-1, Type I receptor binding
interleukin-1, Type II receptor binding
interleukin-1 receptor antagonist activity
interleukin-1 Type I receptor antagonist activity
interleukin-1 Type II receptor antagonist activity

MicroArray Probes:

ABI

Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 778297 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-z4mpm:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0