This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00446735_m1
See other NME7 GE Assays ›
Gene Symbol:
Nme7
Gene Name:
NME/NM23 family member 7
Gene Aliases:
D530024H21Rik, Nm23-M7, Nm23-r7
Chromosome Location:
Chr.1: 164307662 - 164437300 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00446735_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nme7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001293313.1 NP_001280242.1 4 - 5 456 79
NM_138314.4 NP_612187.2 4 - 5 459 79
NM_178071.6 NP_835172.1 4 - 5 456 79
GenBank mRNA AF202048.1 - 3 - 4 79
AK043515.1 - 4 - 5 443 79
AK085222.1 - 4 - 5 404 79
AK133221.1 - 927 79
AK144915.1 - 4 - 5 460 79
AK164652.1 - 4 - 5 404 79
BC038021.1 - 5 - 6 486 3963844 79
BC049225.1 - 3 - 4 387 3483725 79

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100001502.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Nme7

Panther Classification:

Molecular Function -

kinase transferase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

epithelial cilium movement
nucleoside diphosphate phosphorylation
GTP biosynthetic process
UTP biosynthetic process
CTP biosynthetic process
determination of left/right symmetry
brain development
nucleotide metabolic process
phosphorylation
intraciliary transport
ciliary receptor clustering involved in smoothened signaling pathway
left/right pattern formation
nucleotide binding
molecular_function
nucleoside diphosphate kinase activity
ATP binding
kinase activity
transferase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100012395.mm.1 NM_138314.4 Refseq
Affymetrix Mouse Clariom S Array PSR0100012395.mm.1 NM_178071.6 Refseq
Affymetrix Mouse Clariom S Array PSR0100012395.mm.1 TC0100001502.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100012392.mm.1 TC0100001502.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-ng27p:80/100.66.78.145:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline