This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00446809_m1
See other PPP2R4 GE Assays ›
Gene Symbol:
Ppp2r4
Gene Name:
protein phosphatase 2A activator, regulatory subunit B
Gene Aliases:
2610042B21Rik, C77440, N28142, PR53
Chromosome Location:
Chr.2: 30416050 - 30447807 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00446809_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 330.60
330.6
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppp2r4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_138748.5 NP_620087.3 5 - 6 711 65
GenBank mRNA AK031591.1 - 5 - 6 712 65
AK046288.1 - 5 - 6 515 65
AK049184.1 - 5 - 6 702 65
AK150029.1 - 5 - 6 706 65
AK160980.1 - 5 - 6 566 65
AK195195.1 - 2 - 3 165 65
AY035997.1 - 5 - 6 564 65
BC006626.1 - 5 - 6 627 3158086 65
BC052852.1 - 5 - 6 637 30058414 65

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200000535.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ppp2r4

Panther Classification:

Molecular Function -

protein-binding activity modulator phosphatase activator phosphatase modulator

Gene Ontology Categories:

Function(s) Process(es)

protein peptidyl-prolyl isomerization
metabolic process
mitotic spindle organization in nucleus
negative regulation of phosphoprotein phosphatase activity
positive regulation of phosphoprotein phosphatase activity
regulation of protein phosphatase type 2A activity
positive regulation of protein dephosphorylation
negative regulation of protein dephosphorylation
positive regulation of apoptotic process
positive regulation of catalytic activity
regulation of phosphoprotein phosphatase activity
nucleotide binding
peptidyl-prolyl cis-trans isomerase activity
receptor binding
ATP binding
protein tyrosine phosphatase activator activity
protein phosphatase type 2A regulator activity
isomerase activity
ATPase activity
phosphatase activator activity
protein homodimerization activity
protein phosphatase 2A binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200004341.mm.1 NM_138748.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200004341.mm.1 NM_138748.5 Refseq
Affymetrix Mouse Clariom S Array PSR0200004341.mm.1 TC0200000535.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-vvsgg:80/100.66.72.198:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0