This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00446810_g1
See other PPP2R4 GE Assays ›
Gene Symbol:
Ppp2r4
Gene Name:
protein phosphatase 2A activator, regulatory subunit B
Gene Aliases:
2610042B21Rik, C77440, N28142, PR53
Chromosome Location:
Chr.2: 30416050 - 30447807 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00446810_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppp2r4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_138748.5 NP_620087.3 6 - 7 810 68
GenBank mRNA AK031591.1 - 6 - 7 811 68
AK046288.1 - 6 - 7 614 68
AK049184.1 - 6 - 7 801 68
AK150029.1 - 6 - 7 805 68
AK160980.1 - 6 - 7 665 68
BC006626.1 - 6 - 7 726 3158086 68
BC052852.1 - 6 - 7 736 30058414 68

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200000535.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ppp2r4

Panther Classification:

Molecular Function -

protein-binding activity modulator phosphatase activator phosphatase modulator

Gene Ontology Categories:

Function(s) Process(es)

protein peptidyl-prolyl isomerization
metabolic process
mitotic spindle organization in nucleus
negative regulation of phosphoprotein phosphatase activity
positive regulation of phosphoprotein phosphatase activity
regulation of protein phosphatase type 2A activity
positive regulation of protein dephosphorylation
negative regulation of protein dephosphorylation
positive regulation of apoptotic process
positive regulation of catalytic activity
regulation of phosphoprotein phosphatase activity
nucleotide binding
peptidyl-prolyl cis-trans isomerase activity
receptor binding
ATP binding
protein tyrosine phosphatase activator activity
protein phosphatase type 2A regulator activity
isomerase activity
ATPase activity
phosphatase activator activity
protein homodimerization activity
protein phosphatase 2A binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200004345.mm.1 NM_138748.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200004345.mm.1 NM_138748.5 Refseq
Affymetrix Mouse Clariom S Array PSR0200004345.mm.1 TC0200000535.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200004343.mm.1 NM_138748.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200004343.mm.1 NM_138748.5 Refseq
Affymetrix Mouse Clariom S Array PSR0200004343.mm.1 TC0200000535.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline