Assay Details
| Gene Symbol: Plg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008877.3 | NP_032903.3 | 1 - 2 | 110 | 93 | |
| GenBank mRNA | AK040840.1 | - | 1 - 2 | 61 | 93 | |
| AK132254.1 | - | 1 - 2 | 79 | 93 | ||
| BC014773.1 | - | 1 - 2 | 111 | 4193957 | 93 | |
| BC057186.1 | - | 1 - 2 | 85 | 5097123 | 93 | |
| BY131661.1 | - | 1 - 2 | 110 | 93 | ||
| J04766.1 | - | 1 - 2 | 75 | 93 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK132254.1,BC014773.1,BC057186.1,J04766.1,NM_008877.3 transcripts.
Set Membership: |
|

Plg
Genomic Map