This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00447379_g1
See other SSR4 GE Assays ›
Gene Symbol:
Ssr4
Gene Name:
signal sequence receptor, delta
Gene Aliases:
AW743063, C78062, Trap
Chromosome Location:
Chr.X: 73787028 - 73790828 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00447379_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ssr4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001166480.1 NP_001159952.1 3 - 4 475 64
NM_009279.3 NP_033305.1 3 - 4 357 64
GenBank mRNA AK003044.1 - 3 - 4 337 64
AK007921.1 - 3 - 4 313 64
AK013723.1 - 3 - 4 319 64
AK160089.1 - 3 - 4 323 64
AK191259.1 - 1 - 2 57 64
BC008172.1 - 3 - 4 278 3481948 64
BC019419.1 - 2 - 3 1808 64
BI989805.1 - 3 - 4 475 64
BQ747160.1 - 3 - 4 260 64
CJ144362.1 - 3 - 4 64
X90582.1 - 3 - 4 438 64

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0X00000695.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ssr4

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0X00004571.mm.1 NM_001166480.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00004571.mm.1 NM_001166480.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00004571.mm.1 NM_009279.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00004571.mm.1 NM_009279.3 Refseq
Affymetrix Mouse Clariom S Array PSR0X00004571.mm.1 TC0X00000695.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0