This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00447381_g1
See other SSR4 GE Assays ›
Gene Symbol:
Ssr4
Gene Name:
signal sequence receptor, delta
Gene Aliases:
AW743063, C78062, Trap
Chromosome Location:
Chr.X: 73787028 - 73790828 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00447381_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ssr4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001166480.1 NP_001159952.1 5 - 6 626 57
NM_009279.3 NP_033305.1 5 - 6 508 57
GenBank mRNA AK007921.1 - 5 - 6 464 57
AK013723.1 - 5 - 6 470 57
AK160089.1 - 5 - 6 474 57
AK207479.1 - 1 - 2 43 57
AK214704.1 - 1 - 2 43 57
BC008172.1 - 5 - 6 429 3481948 57
BC012971.1 - 2 - 3 2403 57
BC019419.1 - 4 - 5 1959 57
BQ747160.1 - 5 - 6 411 57
X90582.1 - 5 - 6 589 57

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0X00000695.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ssr4

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32464770 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P546090 X90582.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P546090 BC012971.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P546090 BC019419.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P546090 BQ747160.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P546090 AK142600.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P546090 X90582.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P546090 BC012971.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P546090 BC019419.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P546090 BQ747160.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P546090 AK142600.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 338486 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 783148 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 853067 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P127239 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P546090 X90582.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P546090 BC012971.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P546090 BC019419.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P546090 BQ747160.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P546090 AK142600.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P546090 X90582.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P546090 BC012971.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P546090 BC019419.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P546090 BQ747160.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P546090 AK142600.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P174463 X90582.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P174463 BC019419.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P174463 BQ747160.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P174463 AK160089.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P174463 BC008172.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32464777 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32464773 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32464786 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32464785 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32464795 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32464803 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32464784 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32464772 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32464791 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32464775 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32464794 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32464783 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32464792 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32464793 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32464779 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32464814 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32464797 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32464776 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32464815 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32464799 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32464780 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32464806 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32464771 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32464789 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32464802 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P127239 - -
Whole Mouse Genome G4122A A_51_P174463 X90582.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P174463 BC019419.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P174463 BQ747160.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P174463 AK160089.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P174463 BC008172.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P546090 X90582.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P546090 BC012971.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P546090 BC019419.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P546090 BQ747160.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P546090 AK142600.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P546090 X90582.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P546090 BC012971.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P546090 BC019419.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P546090 BQ747160.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P546090 AK142600.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P140003 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P127239 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15159735 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15159692 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32464770 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32464794 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32464786 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32464789 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32464803 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32464785 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32464768 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32464775 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32464779 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32464802 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32464795 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32464777 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32464804 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32464773 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32464784 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32464771 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32464811 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32464797 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32464791 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32464772 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32464799 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32464793 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32464783 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32464806 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P15159719 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32464771 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32464792 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32464773 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32464794 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32464775 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32464804 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32464779 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32464789 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32464802 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32464797 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32464786 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P15035721 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32464780 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P15159701 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32464795 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32464777 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32464783 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32464811 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32464784 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32464799 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32464791 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P15159717 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10600303 - -
Affymetrix Mouse Gene 1.0 ST Array 10600304 - -
Affymetrix Mouse Gene 1.0 ST Array 10600307 - -
Affymetrix Mouse Gene 1.0 ST Array 10600305 - -
Affymetrix Mouse Gene 1.0 ST Array 10600302 - -
Affymetrix Mouse Gene 1.0 ST Array 10600306 - -
Affymetrix HT MG-430 PM Array Plate 1448524_PM_s_at - -
Affymetrix HT MG-430 PM Array Plate 1427096_PM_s_at - -
Affymetrix Mouse Gene 1.1 ST Array 10600301 - -
Mouse Genome 430 2.0 Array 1448524_s_at - -
Mouse Genome 430 2.0 Array 1427096_s_at - -
Mouse Genome 430 2.0 Array 1435709_PM_at - -
Mouse Genome 430 2.0 Array 1435709_at - -
Mouse Genome 430 2.0 Array 1448524_PM_s_at - -
Mouse Genome 430 2.0 Array 1427096_PM_s_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 20832_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 20832_PM_at - -
Affymetrix Mouse Clariom S Array PSR0X00004577.mm.1 NM_001166480.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00004577.mm.1 NM_001166480.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00004577.mm.1 NM_009279.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00004577.mm.1 NM_009279.3 Refseq
Affymetrix Mouse Clariom S Array PSR0X00004577.mm.1 TC0X00000695.mm.1 MTA TC
Mouse Expression Set 430 1448524_PM_s_at - -
Mouse Expression Set 430 1435709_PM_at - -
Mouse Expression Set 430 1448524_s_at - -
Mouse Expression Set 430 1435709_at - -
Mouse Expression Set 430 1427096_s_at - -
Mouse Expression Set 430 1427096_PM_s_at - -
Murine Genome U74v2 Set 101511_at - -
Murine Genome U74v2 Set 101511_PM_at - -
Mouse Genome 430A 2.0 Array 1435709_at - -
Mouse Genome 430A 2.0 Array 1448524_s_at - -
Mouse Genome 430A 2.0 Array 1427096_PM_s_at - -
Mouse Genome 430A 2.0 Array 1448524_PM_s_at - -
Mouse Genome 430A 2.0 Array 1427096_s_at - -
Mouse Genome 430A 2.0 Array 1435709_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1427096_PM_s_at - -
Affymetrix HT Mouse Genome 430A Array 1435709_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1427096_s_at - -
Affymetrix HT Mouse Genome 430A Array 1448524_s_at - -
Affymetrix HT Mouse Genome 430A Array 1448524_PM_s_at - -
Affymetrix HT Mouse Genome 430A Array 1435709_at - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 4150132 - -
Illumina mouse-6 v1.0 expression beadchip 4150132 - -
Illumina mouse-6 v1.0 expression beadchip 1340619 - -
Illumina mouseRef-8 v1.0 expression beadchip 0004150132 - -
Illumina mouseRef-8 v1.0 expression beadchip 0001340619 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2773447 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_1255237 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1255237 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2773447 - -
mouse-6 v1.1 (Illumina) scl54843.2_132-S - -
mouse-6 v1.1 (Illumina) scl0020832.1_299-S - -
Illumina Sentrix mouse-6.v1 bead array 1340619 - -
Illumina Sentrix mouse-6.v1 bead array 4150132 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl54843.2_132-S - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl0020832.1_299-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 1340619 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 4150132 - -
MouseRef-8 v2 Expression BeadChip ILMN_1255237 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2773447 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_1255237 - -
Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0020832.1_299-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (3) scl54843.2_132-S - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0