This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00448202_m1
See other PLTP GE Assays ›
Gene Symbol:
Pltp
Gene Name:
phospholipid transfer protein
Gene Aliases:
Bpife, OD107
Chromosome Location:
Chr.2: 164839518 - 164857708 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00448202_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pltp
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011125.2 NP_035255.1 8 - 9 841 70
GenBank mRNA AK148330.1 - 8 - 9 846 70
AK149730.1 - 8 - 9 772 70
U37226.1 - 8 - 9 771 70

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200005136.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pltp

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
transport
lipid transport
vitamin E biosynthetic process
positive regulation of cholesterol efflux
sperm motility
high-density lipoprotein particle remodeling
lipid binding

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21820156 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21820159 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21820158 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21820155 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21820157 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21820163 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21820161 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1993955 BC018534.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1993955 J05261.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1993955 AK032197.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1993955 AK159150.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1993955 NM_008906.3 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 683211 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 400243 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 16734 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 659342 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 855005 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 834439 XM_001475790.1 refseq_mrna_mouse_XM
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 834439 XM_001477715.1 refseq_mrna_mouse_XM
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 73637 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 691687 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 283411 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 396470 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 179807 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 24681 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P175564 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P110325 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1993955 BC018534.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1993955 J05261.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1993955 AK032197.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1993955 AK159150.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1993955 NM_008906.3 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P226655 U28960.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P226655 U37226.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P226655 AK148330.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P226655 BC003782.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P226655 CT010317.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21820167 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21820184 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21820186 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21820163 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21820164 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21820187 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21820148 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21820180 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21820157 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21820162 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21820160 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21820159 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21820169 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21820174 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21820151 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21820179 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21820153 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21820154 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21820170 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21820188 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21820165 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21820175 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21820177 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21820172 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P175564 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P110325 - -
Whole Mouse Genome G4122A A_51_P226655 U28960.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P226655 U37226.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P226655 AK148330.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P226655 BC003782.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P226655 CT010317.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P125215 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P110325 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P175564 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01734927 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01734929 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01734926 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01734915 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01835569 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21820163 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21820157 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21820159 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21820155 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21820156 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21820158 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21820161 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820033 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820097 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820003 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820035 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820055 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820202 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820085 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820184 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820063 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820163 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820094 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820200 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820021 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820157 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820111 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820132 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820044 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820104 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820125 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820026 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820169 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820088 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820028 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820012 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820134 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820015 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819999 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820153 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820050 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820162 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820121 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820095 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820165 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820031 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820009 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820056 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820048 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820160 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820066 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820175 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820179 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820054 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820030 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820128 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820073 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820046 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820068 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820119 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820186 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820180 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820177 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820148 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820059 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820074 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820039 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820137 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820078 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820106 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820171 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820032 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820120 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820167 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820159 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820143 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820112 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820142 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820124 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820139 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820115 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820076 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820017 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820154 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820069 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820023 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820172 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820040 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820187 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820196 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820192 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820001 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820114 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21820170 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21820134 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21820137 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01338930 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01734915 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01734918 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21820132 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21820193 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02071084 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21820169 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01648582 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01734909 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21820189 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01403905 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21820186 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01872478 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21820144 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21820148 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21820100 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21820135 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02222606 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21820142 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21820101 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21820184 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21820175 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01734907 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01734916 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01734911 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01391916 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21820106 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01648583 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21820128 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21820141 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21820110 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01872477 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21820129 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21820153 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21820109 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21820165 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21820171 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21820191 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21820139 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21820177 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01391921 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01987184 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02251964 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21820126 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21820187 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01734913 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01213652 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21820172 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10489580 - -
Affymetrix Mouse Gene 1.0 ST Array 10489572 - -
Affymetrix Mouse Gene 1.0 ST Array 10489578 - -
Affymetrix Mouse Gene 1.0 ST Array 10489581 - -
Affymetrix Mouse Gene 1.0 ST Array 10489582 - -
Affymetrix Mouse Gene 1.0 ST Array 10489575 - -
Affymetrix Mouse Gene 1.0 ST Array 10489577 - -
Affymetrix Mouse Gene 1.0 ST Array 10489571 - -
Affymetrix Mouse Gene 1.0 ST Array 10489584 - -
Affymetrix Mouse Gene 1.0 ST Array 10489570 - -
Affymetrix Mouse Gene 1.0 ST Array 10489576 - -
Affymetrix Mouse Gene 1.0 ST Array 10489579 - -
Affymetrix Mouse Gene 1.0 ST Array 10489573 - -
Affymetrix Mouse Gene 1.0 ST Array 10489583 - -
Affymetrix Mouse Gene 1.0 ST Array 10489574 - -
Affymetrix Mouse Gene 1.0 ST Array 10489586 - -
Affymetrix Mouse Gene 1.0 ST Array 10489587 - -
Affymetrix Mouse Gene 1.0 ST Array 10489588 - -
Affymetrix Mouse Gene 1.0 ST Array 10489585 - -
Affymetrix HT MG-430 PM Array Plate 1417963_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1456424_PM_s_at - -
Affymetrix Mouse Gene 1.1 ST Array 10489569 - -
Mouse Genome 430 2.0 Array 1456424_PM_s_at - -
Mouse Genome 430 2.0 Array 1445659_at - -
Mouse Genome 430 2.0 Array 1456424_s_at - -
Mouse Genome 430 2.0 Array 1417963_at - -
Mouse Genome 430 2.0 Array 1445659_PM_at - -
Mouse Genome 430 2.0 Array 1417963_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 18830_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 18830_at - -
Affymetrix Murine Genome 6500A Array Msa17220_at - -
Affymetrix Mouse Clariom S Array PSR0200043783.mm.1 NM_011125.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200043783.mm.1 NM_011125.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200043783.mm.1 TC0200005136.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200043781.mm.1 NM_011125.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200043781.mm.1 NM_011125.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200043781.mm.1 TC0200005136.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200043782.mm.1 NM_011125.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200043782.mm.1 NM_011125.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200043782.mm.1 TC0200005136.mm.1 MTA TC
Mouse Expression Set 430 1417963_PM_at - -
Mouse Expression Set 430 1456424_PM_s_at - -
Mouse Expression Set 430 1456424_s_at - -
Mouse Expression Set 430 1417963_at - -
Murine Genome U74v2 Set 100927_PM_at - -
Murine Genome U74v2 Set 100927_at - -
Mouse Genome 430A 2.0 Array 1417963_at - -
Mouse Genome 430A 2.0 Array 1456424_s_at - -
Mouse Genome 430A 2.0 Array 1417963_PM_at - -
Mouse Genome 430A 2.0 Array 1456424_PM_s_at - -
Affymetrix HT Mouse Genome 430A Array 1456424_PM_s_at - -
Affymetrix HT Mouse Genome 430A Array 1417963_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1417963_at - -
Affymetrix HT Mouse Genome 430A Array 1456424_s_at - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 130497 - -
Illumina mouse-6 v1.0 expression beadchip 4050139 - -
Illumina mouse-6 v1.0 expression beadchip 130497 - -
Illumina mouse-6 v1.0 expression beadchip 630110 - -
Illumina mouseRef-8 v1.0 expression beadchip 0000630110 - -
Illumina mouseRef-8 v1.0 expression beadchip 0000130497 - -
Illumina mouseRef-8 v1.0 expression beadchip 0004050139 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2663014 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2589401 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2663014 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2589401 - -
mouse-6 v1.1 (Illumina) scl0003032.1_86-S - -
mouse-6 v1.1 (Illumina) scl18344.17.1_25-S - -
mouse-6 v1.1 (Illumina) scl0003117.1_12-S - -
Illumina Sentrix mouse-6.v1 bead array 4050139 - -
Illumina Sentrix mouse-6.v1 bead array 130497 - -
Illumina Sentrix mouse-6.v1 bead array 630110 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl0003117.1_12-S - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl0003032.1_86-S - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl18344.17.1_25-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 630110 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 130497 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 4050139 - -
MouseRef-8 v2 Expression BeadChip ILMN_2669714 - -
MouseRef-8 v2 Expression BeadChip ILMN_2589401 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2669714 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2589401 - -
Illumina Sentrix Mouse-6 Expression BeadChip (3) scl18344.17.1_25-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0003032.1_86-S - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline