This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00448212_g1
See other PNLIPRP2 GE Assays ›
Gene Symbol:
Pnliprp2
Gene Name:
pancreatic lipase-related protein 2
Gene Aliases:
PLRP2
Chromosome Location:
Chr.19: 58759723 - 58777534 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00448212_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pnliprp2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011128.2 NP_035258.2 3 - 4 432 78
GenBank mRNA AK008021.1 - 3 - 4 437 78
AK131882.1 - 3 - 4 418 78
BC094923.1 - 3 - 4 391 6431510 78
M30687.1 - 3 - 4 425 78

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1900000843.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pnliprp2

Panther Classification:

Molecular Function -

lipase

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
cellular defense response
digestion
phospholipid catabolic process
lipid catabolic process
galactolipid catabolic process
phospholipase activity
triglyceride lipase activity
calcium ion binding
lipase activity
hydrolase activity
metal ion binding
acylglycerol lipase activity
galactolipase activity
carboxylic ester hydrolase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1900007317.mm.1 NM_011128.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900007317.mm.1 NM_011128.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900007317.mm.1 TC1900000843.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-cjlz8:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0