This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00448436_g1
See other PTPN11 GE Assays ›
Gene Symbol:
Ptpn11
Gene Name:
protein tyrosine phosphatase, non-receptor type 11
Gene Aliases:
2700084A17Rik, AW536184, PTP1D, PTP2C, SAP-2, SH-PTP2, SH-PTP3, SHP-2, Shp2, Syp
Chromosome Location:
Chr.5: 121130533 - 121191397 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00448436_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ptpn11
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001109992.1 NP_001103462.1 5 - 6 824 62
NM_011202.3 NP_035332.1 5 - 6 824 62
GenBank mRNA AK011468.1 - 5 - 6 786 62
AK147354.1 - 5 - 6 795 62
AK159501.1 - 5 - 6 824 62
AK159587.1 - 5 - 6 824 62
AK163809.1 - 2 - 3 252 62
BC028513.1 - 5 - 6 774 62
BC057398.1 - 5 - 6 756 6409640 62
BC059278.1 - 5 - 6 802 2866038 62
D84372.1 - 5 - 6 700 62

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500003184.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Protein Phosphatase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ptpn11

Panther Classification:

Molecular Function -

protein phosphatase

Gene Ontology Categories:

Function(s) Process(es)

DNA damage checkpoint
activation of MAPK activity
protein dephosphorylation
lipid metabolic process
triglyceride metabolic process
epidermal growth factor receptor signaling pathway
integrin-mediated signaling pathway
axonogenesis
brain development
heart development
hormone-mediated signaling pathway
positive regulation of signal transduction
dephosphorylation
platelet formation
microvillus organization
abortive mitotic cell cycle
regulation of cell adhesion mediated by integrin
negative regulation of cell adhesion mediated by integrin
multicellular organism growth
organ growth
peptidyl-tyrosine dephosphorylation
megakaryocyte development
atrioventricular canal development
ERBB signaling pathway
regulation of multicellular organism growth
hormone metabolic process
glucose homeostasis
positive regulation of mitotic cell cycle
negative regulation of insulin secretion
regulation of protein export from nucleus
positive regulation of hormone secretion
negative regulation of hormone secretion
platelet-derived growth factor receptor signaling pathway
neurotrophin TRK receptor signaling pathway
ephrin receptor signaling pathway
multicellular organismal reproductive process
genitalia development
inner ear development
homeostasis of number of cells within a tissue
negative regulation of cortisol secretion
negative regulation of growth hormone secretion
face morphogenesis
intestinal epithelial cell migration
positive regulation of ERK1 and ERK2 cascade
positive regulation of glucose import in response to insulin stimulus
phosphoprotein phosphatase activity
protein tyrosine phosphatase activity
non-membrane spanning protein tyrosine phosphatase activity
SH3/SH2 adaptor activity
insulin receptor binding
protein binding
hydrolase activity
phosphatase activity
protein domain specific binding
receptor tyrosine kinase binding
D1 dopamine receptor binding
phospholipase binding
insulin receptor substrate binding
peptide hormone receptor binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500028330.mm.1 TC0500003184.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500028330.mm.1 NM_001109992.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500028330.mm.1 NM_001109992.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500028330.mm.1 NM_011202.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500028330.mm.1 NM_011202.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7dtsd:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0