This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00448541_g1
See other RAD52 GE Assays ›
Gene Symbol:
Rad52
Gene Name:
RAD52 homolog (S. cerevisiae)
Gene Aliases:
Rad52yh
Chromosome Location:
Chr.6: 119902207 - 119922823 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00448541_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rad52
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001166381.1 NP_001159853.1 4 - 5 424 63
NM_001166382.1 NP_001159854.1 4 - 5 424 63
NM_001166383.1 NP_001159855.1 4 - 5 424 63
NM_011236.2 NP_035366.2 4 - 5 424 63
GenBank mRNA AK080702.1 - 4 - 5 397 63
BC019123.1 - 4 - 5 404 5003696 63
U06837.1 - 4 - 5 346 63
U12135.1 - 4 - 5 393 63
Z32767.1 - 4 - 5 418 63

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600001340.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rad52

Panther Classification:

Molecular Function -

DNA metabolism protein nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

double-strand break repair via homologous recombination
DNA repair
DNA recombination
cellular response to DNA damage stimulus
DNA double-strand break processing involved in repair via single-strand annealing
cellular response to oxidative stress
double-strand break repair via single-strand annealing
regulation of nucleotide-excision repair
recombinase activity
DNA binding
identical protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600010642.mm.1 TC0600001340.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600010642.mm.1 NM_001166381.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010642.mm.1 NM_001166381.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010642.mm.1 NM_001166382.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010642.mm.1 NM_001166382.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010642.mm.1 NM_001166383.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010642.mm.1 NM_001166383.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010642.mm.1 NM_011236.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010642.mm.1 NM_011236.2 Refseq
Affymetrix Mouse Clariom S Array PSR0600010643.mm.1 TC0600001340.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600010643.mm.1 NM_001166381.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010643.mm.1 NM_001166381.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010643.mm.1 NM_001166382.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010643.mm.1 NM_001166382.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010643.mm.1 NM_001166383.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010643.mm.1 NM_001166383.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010643.mm.1 NM_011236.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010643.mm.1 NM_011236.2 Refseq
Affymetrix Mouse Clariom S Array PSR0600010645.mm.1 TC0600001340.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600010645.mm.1 NM_001166381.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010645.mm.1 NM_001166381.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010645.mm.1 NM_001166382.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010645.mm.1 NM_001166382.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010645.mm.1 NM_001166383.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010645.mm.1 NM_001166383.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010645.mm.1 NM_011236.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010645.mm.1 NM_011236.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-nbrrd:80/100.66.77.69:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0