This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00448771_m1
See other SMARCB1 GE Assays ›
Gene Symbol:
Smarcb1
Gene Name:
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily b, member 1
Gene Aliases:
AU020204, Baf47, Ini1, SNF5/INI1, Snf5
Chromosome Location:
Chr.10: 75896769 - 75921614 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00448771_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Smarcb1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011418.2 NP_035548.1 2 - 3 454 108
GenBank mRNA AB017343.1 - 2 - 3 235 108
AB041578.1 - 2 - 3 409 108
AJ011739.1 - 2 - 3 428 108
AJ011740.1 - 2 - 3 428 108
AK075906.1 - 2 - 3 437 108
BC025163.1 - 2 - 3 408 3154501 108

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Smarcb1

Panther Classification:

Molecular Function -

DNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

blastocyst development
blastocyst hatching
DNA repair
nucleosome disassembly
chromatin remodeling
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
cell cycle
nervous system development
negative regulation of cell proliferation
chromatin modification
cell differentiation
single stranded viral RNA replication via double stranded DNA intermediate
ATP-dependent chromatin remodeling
positive regulation by host of viral transcription
mitotic cell cycle phase transition
positive regulation of transcription from RNA polymerase II promoter
positive regulation of sequence-specific DNA binding transcription factor activity
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II distal enhancer sequence-specific DNA binding
p53 binding
transcription coactivator activity
protein binding
Tat protein binding
nucleosomal DNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-7669c5b596-s62d2:80/100.66.75.204:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0