This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00448964_m1
See other TFEB GE Assays ›
Gene Symbol:
Tfeb
Gene Name:
transcription factor EB
Gene Aliases:
Tcfeb, bHLHe35
Chromosome Location:
Chr.17: 47737037 - 47792416 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00448964_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tfeb
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001161722.1 NP_001155194.1 4 - 5 853 79
NM_001161723.1 NP_001155195.1 4 - 5 847 79
NM_011549.3 NP_035679.3 3 - 4 794 79
GenBank mRNA AF079095.1 - 3 - 4 775 79
AK078755.1 - 4 - 5 785 79
AK154971.1 - 4 - 5 851 79
BC064789.1 - 3 - 4 794 30429545 79

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1700000924.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tfeb

Gene Ontology Categories:

Function(s) Process(es)

embryonic placenta development
immune system process
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
autophagy
humoral immune response
lysosome organization
positive regulation of autophagy
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
transcription factor activity, sequence-specific DNA binding
transcription regulatory region DNA binding
protein heterodimerization activity
protein dimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-4sktr:80/100.66.76.202:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0