This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00449002_m1
See other TEAD2 GE Assays ›
Gene Symbol:
Tead2
Gene Name:
TEA domain family member 2
Gene Aliases:
ETF, Etdf, TEAD-2, TEF-4, TEF4
Chromosome Location:
Chr.7: 45215753 - 45233636 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00449002_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tead2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001285498.1 NP_001272427.1 4 - 5 489 81
NM_001285500.1 NP_001272429.1 4 - 5 342 81
NM_011565.2 NP_035695.1 4 - 5 447 81
GenBank mRNA AK132009.1 - 3 - 4 382 81
AK137064.1 - 3 - 4 1730 81
AK143155.1 - 3 - 4 1663 81
AK144857.1 - 4 - 5 409 81
AK146595.1 - 4 - 5 409 81
BC050217.1 - 3 - 4 267 30021393 81
D50563.1 - 4 - 5 447 81
X94442.1 - 3 - 4 361 81
Y10026.1 - 4 - 5 411 81

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700000835.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tead2

Panther Classification:

Molecular Function -

DNA-binding transcription factor

Gene Ontology Categories:

Function(s) Process(es)

vasculogenesis
neural tube closure
embryonic heart tube morphogenesis
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
notochord development
hippo signaling
positive regulation of transcription from RNA polymerase II promoter
paraxial mesoderm development
lateral mesoderm development
negative regulation of cell death
cellular response to retinoic acid
regulation of stem cell differentiation
DNA binding
transcription factor activity, sequence-specific DNA binding
transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700007164.mm.1 TC0700000835.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700007164.mm.1 NM_001285498.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700007164.mm.1 NM_001285500.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700007164.mm.1 NM_011565.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700007164.mm.1 NM_011565.2 Refseq
Affymetrix Mouse Clariom S Array PSR0700007168.mm.1 TC0700000835.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700007168.mm.1 NM_001285498.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700007168.mm.1 NM_001285500.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700007168.mm.1 NM_011565.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700007168.mm.1 NM_011565.2 Refseq
Affymetrix Mouse Clariom S Array PSR0700007165.mm.1 TC0700000835.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700007165.mm.1 NM_001285498.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700007165.mm.1 NM_001285500.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700007165.mm.1 NM_011565.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700007165.mm.1 NM_011565.2 Refseq
Affymetrix Mouse Clariom S Array PSR0700007162.mm.1 TC0700000835.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700007162.mm.1 NM_001285498.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700007162.mm.1 NM_001285500.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700007162.mm.1 NM_011565.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700007162.mm.1 NM_011565.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0