This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00450173_m1
See other GNE GE Assays ›
Gene Symbol:
Gne
Gene Name:
glucosamine (UDP-N-acetyl)-2-epimerase/N-acetylmannosamine kinase
Gene Aliases:
2310066H07Rik, DMRV, GLCNE, IBM2, NM, Uae1
Chromosome Location:
Chr.4: 44034075 - 44084177 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00450173_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gne
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001190414.1 NP_001177343.1 4 - 5 1125 64
NM_015828.3 NP_056643.3 4 - 5 943 64
GenBank mRNA AJ132236.1 - 3 - 4 771 64
AK136573.1 - 4 - 5 944 64
AK170707.1 - 4 - 5 1126 64
AM697710.1 - 4 - 5 864 64
BC015277.1 - 4 - 5 931 4216299 64
BC051254.1 - 4 - 5 931 6478532 64

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400002549.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gne

Panther Classification:

Molecular Function -

DNA-binding transcription factor

Gene Ontology Categories:

Function(s) Process(es)

carbohydrate metabolic process
UDP-N-acetylglucosamine metabolic process
N-acetylneuraminate metabolic process
metabolic process
phosphorylation
carbohydrate phosphorylation
nucleotide binding
catalytic activity
hydrolase activity, hydrolyzing O-glycosyl compounds
ATP binding
UDP-N-acetylglucosamine 2-epimerase activity
N-acylmannosamine kinase activity
kinase activity
transferase activity
phosphotransferase activity, alcohol group as acceptor
hydrolase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400020806.mm.1 TC0400002549.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400020806.mm.1 NM_001190414.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400020806.mm.1 NM_001190414.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400020806.mm.1 NM_015828.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400020806.mm.1 NM_015828.3 Refseq
Affymetrix Mouse Clariom S Array PSR0400020807.mm.1 TC0400002549.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400020807.mm.1 NM_001190414.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400020807.mm.1 NM_001190414.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400020807.mm.1 NM_015828.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400020807.mm.1 NM_015828.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0