This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00450304_g1
See other TNK2 GE Assays ›
Gene Symbol:
Tnk2
Gene Name:
tyrosine kinase, non-receptor, 2
Gene Aliases:
Ack, Ack-1, Ack1, Cdgip, Pyk1
Chromosome Location:
Chr.16: 32644099 - 32683493 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00450304_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tnk2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001110147.1 NP_001103617.1 5 - 6 1098 69
NM_016788.3 NP_058068.2 5 - 6 1098 69
GenBank mRNA AF037260.1 - 5 - 6 881 69
AK087965.1 - 2 - 3 255 69
BC052421.1 - 5 - 6 1098 6400209 69
BU702226.1 - 1 - 2 80 69
DQ666696.1 - 5 - 6 909 69

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tnk2

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
endocytosis
signal transduction
transmembrane receptor protein tyrosine kinase signaling pathway
phosphorylation
cell migration
peptidyl-tyrosine phosphorylation
cell differentiation
peptidyl-tyrosine autophosphorylation
regulation of cell proliferation
innate immune response
positive regulation of peptidyl-tyrosine phosphorylation
regulation of clathrin-mediated endocytosis
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
protein serine/threonine/tyrosine kinase activity
protein tyrosine kinase activity
non-membrane spanning protein tyrosine kinase activity
epidermal growth factor receptor binding
protein binding
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
identical protein binding
metal ion binding
WW domain binding
hormone receptor binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-vh84l:80/100.66.79.51:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0