This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00450567_m1
See other KAT6B GE Assays ›
Gene Symbol:
Kat6b
Gene Name:
K(lysine) acetyltransferase 6B
Gene Aliases:
AI507552, B130044K16Rik, MYST-4, Morf, Myst4, mKIAA0383, qkf, querkopf
Chromosome Location:
Chr.14: 21495310 - 21672478 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00450567_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Kat6b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001205241.1 NP_001192170.1 6 - 7 1368 79
NM_017479.3 NP_059507.2 6 - 7 1492 79
GenBank mRNA AF222800.1 - 6 - 7 1408 79
AK045188.1 - 6 - 7 1504 79
AK147509.1 - 6 - 7 1369 79
BC095974.1 - 5 - 6 1145 79

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1400000195.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Kat6b

Panther Classification:

Molecular Function -

histone modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

nucleosome assembly
transcription, DNA-templated
regulation of transcription, DNA-templated
chromatin modification
histone acetylation
positive regulation of transcription from RNA polymerase II promoter
DNA binding
histone acetyltransferase activity
zinc ion binding
transferase activity
transferase activity, transferring acyl groups
transferase activity, transferring acyl groups other than amino-acyl groups
protein complex binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400001528.mm.1 NM_001205241.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400001528.mm.1 NM_001205241.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400001528.mm.1 NM_017479.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400001528.mm.1 NM_017479.3 Refseq
Affymetrix Mouse Clariom S Array PSR1400001528.mm.1 TC1400000195.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0