This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00450898_g1
See other PQBP1 GE Assays ›
Gene Symbol:
Pqbp1
Gene Name:
polyglutamine binding protein 1
Gene Aliases:
PQBP-1, Sfc2, npw38
Chromosome Location:
Chr.X: 7894519 - 7899269 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00450898_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pqbp1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001252528.1 NP_001239457.1 4 - 5 341 62
NM_001252529.1 NP_001239458.1 4 - 5 407 62
NM_019478.4 NP_062351.2 4 - 5 536 62
GenBank mRNA AJ250406.1 - 3 - 4 287 62
AK077652.1 - 4 - 5 366 62
BC009657.1 - 3 - 4 309 3484619 62
BC051673.1 - 4 - 5 346 30015428 62
BC157976.1 - 4 - 5 441 9088137 62
BQ550428.1 - 4 - 5 412 62
CJ143105.1 - 4 - 5 342 62
CN718083.1 - 3 - 4 321 62

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0X00001784.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pqbp1

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

alternative mRNA splicing, via spliceosome
transcription, DNA-templated
regulation of transcription, DNA-templated
mRNA processing
RNA splicing
neuron projection development
stress granule assembly
regulation of RNA splicing
regulation of dendrite morphogenesis
cilium morphogenesis
protein C-terminus binding
ribonucleoprotein complex binding

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32240780 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1971594 AK077652.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1971594 BC009657.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1971594 BC051673.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1971594 NM_019478.3 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2070254 AK077652.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2070254 BC009657.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2070254 BC051673.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2070254 BC157976.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2070254 NM_019478.3 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2070250 BC157976.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 266637 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 171651 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 717461 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 647798 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P101557 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2070254 AK077652.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2070254 BC009657.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2070254 BC051673.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2070254 BC157976.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2070254 NM_019478.3 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1971594 AK077652.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1971594 BC009657.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1971594 BC051673.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1971594 NM_019478.3 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P135322 AJ250406.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240754 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240766 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240771 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240768 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240752 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240757 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240778 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240748 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240764 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240769 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240767 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240775 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240763 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240772 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240777 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240762 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240759 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240765 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240750 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240773 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240780 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240776 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240755 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32240774 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09227085 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P101557 - -
Whole Mouse Genome G4122A A_51_P135322 AJ250406.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P112808 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P101557 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14941581 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14941590 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32240780 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240699 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240722 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240720 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240716 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240759 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240703 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240701 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240774 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240757 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240776 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240756 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240772 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240750 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240763 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240711 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240771 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240766 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240714 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240706 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240767 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240753 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240780 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240721 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240705 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240709 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240696 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240778 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240708 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240764 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240702 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240762 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240768 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240755 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240704 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240775 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240765 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240697 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32240694 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14534934 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32240764 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32240776 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14772639 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32240753 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32240767 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14517909 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32240777 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14772636 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32240757 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32240769 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32240774 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32240755 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32240750 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14517913 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32240765 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32240756 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32240762 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14517915 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32240768 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32240770 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14941581 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10603379 - -
Affymetrix Mouse Gene 1.0 ST Array 10603375 - -
Affymetrix Mouse Gene 1.0 ST Array 10603374 - -
Affymetrix Mouse Gene 1.0 ST Array 10603378 - -
Affymetrix Mouse Gene 1.0 ST Array 10603380 - -
Affymetrix Mouse Gene 1.0 ST Array 10603377 - -
Affymetrix Mouse Gene 1.0 ST Array 10603381 - -
Affymetrix HT MG-430 PM Array Plate 1417731_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10603373 - -
Mouse Genome 430 2.0 Array 1417731_PM_at - -
Mouse Genome 430 2.0 Array 1417731_at - -
Mouse Genome 430 2.0 Array 1456086_x_at - -
Mouse Genome 430 2.0 Array 1456086_PM_x_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 54633_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 54633_at - -
Affymetrix Mouse Clariom S Array PSR0X00011291.mm.1 NM_001252528.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00011291.mm.1 NM_001252528.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00011291.mm.1 NM_001252529.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00011291.mm.1 NM_001252529.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00011291.mm.1 NM_019478.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00011291.mm.1 NM_019478.4 Refseq
Affymetrix Mouse Clariom S Array PSR0X00011291.mm.1 TC0X00001784.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0X00011292.mm.1 NM_001252528.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00011292.mm.1 NM_001252528.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00011292.mm.1 NM_001252529.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00011292.mm.1 NM_001252529.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00011292.mm.1 NM_019478.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00011292.mm.1 NM_019478.4 Refseq
Affymetrix Mouse Clariom S Array PSR0X00011292.mm.1 TC0X00001784.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0X00011297.mm.1 NM_001252528.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00011297.mm.1 NM_001252528.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00011297.mm.1 NM_001252529.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00011297.mm.1 NM_001252529.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00011297.mm.1 NM_019478.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00011297.mm.1 NM_019478.4 Refseq
Affymetrix Mouse Clariom S Array PSR0X00011297.mm.1 TC0X00001784.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0X00011296.mm.1 NM_001252528.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00011296.mm.1 NM_001252528.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00011296.mm.1 NM_001252529.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00011296.mm.1 NM_001252529.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00011296.mm.1 NM_019478.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00011296.mm.1 NM_019478.4 Refseq
Affymetrix Mouse Clariom S Array PSR0X00011296.mm.1 TC0X00001784.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0X00011293.mm.1 NM_001252528.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00011293.mm.1 NM_001252528.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00011293.mm.1 NM_001252529.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00011293.mm.1 NM_001252529.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00011293.mm.1 NM_019478.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00011293.mm.1 NM_019478.4 Refseq
Affymetrix Mouse Clariom S Array PSR0X00011293.mm.1 TC0X00001784.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0X00011294.mm.1 NM_001252528.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00011294.mm.1 NM_001252528.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00011294.mm.1 NM_001252529.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00011294.mm.1 NM_001252529.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00011294.mm.1 NM_019478.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00011294.mm.1 NM_019478.4 Refseq
Affymetrix Mouse Clariom S Array PSR0X00011294.mm.1 TC0X00001784.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0X00011295.mm.1 NM_001252528.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00011295.mm.1 NM_001252528.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00011295.mm.1 NM_001252529.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00011295.mm.1 NM_001252529.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00011295.mm.1 NM_019478.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00011295.mm.1 NM_019478.4 Refseq
Affymetrix Mouse Clariom S Array PSR0X00011295.mm.1 TC0X00001784.mm.1 MTA TC
Mouse Expression Set 430 1456086_x_at - -
Mouse Expression Set 430 1417731_PM_at - -
Mouse Expression Set 430 1417731_at - -
Mouse Expression Set 430 1456086_PM_x_at - -
Murine Genome U74v2 Set 93010_PM_at - -
Murine Genome U74v2 Set 93010_at - -
Mouse Genome 430A 2.0 Array 1417731_at - -
Mouse Genome 430A 2.0 Array 1456086_PM_x_at - -
Mouse Genome 430A 2.0 Array 1456086_x_at - -
Mouse Genome 430A 2.0 Array 1417731_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1417731_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1417731_at - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 6520603 - -
Illumina mouse-6 v1.0 expression beadchip 6520603 - -
Illumina mouseRef-8 v1.0 expression beadchip 0006520603 - -
mouse-6 v1.1 (Illumina) scl54442.9.1_77-S - -
Illumina Sentrix mouse-6.v1 bead array 6520603 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl54442.9.1_77-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 6520603 - -
MouseRef-8 v2 Expression BeadChip ILMN_2883110 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2677391 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2883110 - -
Illumina Sentrix Mouse-6 Expression BeadChip (3) scl54442.9.1_77-S - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-gm74s:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline