This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00450995_g1
See other GDE1 GE Assays ›
Gene Symbol:
Gde1
Gene Name:
glycerophosphodiester phosphodiesterase 1
Gene Aliases:
1200003M13Rik, Mir16, RGS16
Chromosome Location:
Chr.7: 118688558 - 118705738 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00450995_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gde1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_019580.4 NP_062526.1 3 - 4 630 123
GenBank mRNA AF212860.1 - 3 - 4 611 123
AK004583.1 - 3 - 4 643 123
AK005361.1 - 3 - 4 631 123
AK150807.1 - 3 - 4 652 123
BC003902.1 - 3 - 4 589 3601420 123

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700004176.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gde1

Panther Classification:

Molecular Function -

phosphodiesterase

Gene Ontology Categories:

Function(s) Process(es)

glycerol metabolic process
lipid metabolic process
G-protein coupled receptor signaling pathway
phosphoric diester hydrolase activity
glycerophosphodiester phosphodiesterase activity
hydrolase activity
metal ion binding
glycerophosphoinositol glycerophosphodiesterase activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700034734.mm.1 NM_019580.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700034734.mm.1 NM_019580.4 Refseq
Affymetrix Mouse Clariom S Array PSR0700034734.mm.1 TC0700004176.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700034732.mm.1 NM_019580.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700034732.mm.1 NM_019580.4 Refseq
Affymetrix Mouse Clariom S Array PSR0700034732.mm.1 TC0700004176.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700034737.mm.1 NM_019580.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700034737.mm.1 NM_019580.4 Refseq
Affymetrix Mouse Clariom S Array PSR0700034737.mm.1 TC0700004176.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700034736.mm.1 NM_019580.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700034736.mm.1 NM_019580.4 Refseq
Affymetrix Mouse Clariom S Array PSR0700034736.mm.1 TC0700004176.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700034733.mm.1 NM_019580.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700034733.mm.1 NM_019580.4 Refseq
Affymetrix Mouse Clariom S Array PSR0700034733.mm.1 TC0700004176.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 569558 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-lcddp:80/100.66.76.150:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0