This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00451008_m1
See other PITPNB GE Assays ›
Gene Symbol:
Pitpnb
Gene Name:
phosphatidylinositol transfer protein, beta
Gene Aliases:
PI-TP-beta
Chromosome Location:
Chr.5: 111330697 - 111388361 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00451008_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pitpnb
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001301643.1 NP_001288572.1 9 - 10 755 74
NM_001301644.1 NP_001288573.1 9 - 10 724 74
NM_001301666.1 NP_001288595.1 8 - 9 671 74
NM_019640.5 NP_062614.1 9 - 10 755 74
GenBank mRNA AK028235.1 - 9 - 10 690 74
BC034676.1 - 9 - 10 645 4481685 74
U46934.1 - 9 - 10 657 74

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500001162.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Transporter 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pitpnb

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

in utero embryonic development
transport
phospholipid transport
phospholipid binding
lipid binding
phosphatidylinositol transporter activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500009969.mm.1 NM_019640.5 Refseq
Affymetrix Mouse Clariom S Array PSR0500009969.mm.1 TC0500001162.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500009968.mm.1 NM_019640.5 Refseq
Affymetrix Mouse Clariom S Array PSR0500009968.mm.1 TC0500001162.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 675715 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline