This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00451729_g1
See other PPP1R1A GE Assays ›
Gene Symbol:
Ppp1r1a
Gene Name:
protein phosphatase 1, regulatory (inhibitor) subunit 1A
Gene Aliases:
0610038N18Rik, I-1
Chromosome Location:
Chr.15: 103530279 - 103538005 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00451729_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppp1r1a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_021391.3 NP_067366.1 3 - 4 303 78
GenBank mRNA AF281676.1 - 3 - 4 193 78
BC025123.1 - 3 - 4 272 4019442 78
CA321941.1 - 3 - 4 303 78

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500002323.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ppp1r1a

Panther Classification:

Molecular Function -

phosphatase inhibitor

Gene Ontology Categories:

Function(s) Process(es)

carbohydrate metabolic process
glycogen metabolic process
negative regulation of protein kinase activity
signal transduction
intracellular signal transduction
protein phosphatase inhibitor activity
protein serine/threonine phosphatase inhibitor activity
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500019290.mm.1 NM_021391.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500019290.mm.1 NM_021391.3 Refseq
Affymetrix Mouse Clariom S Array PSR1500019290.mm.1 TC1500002323.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0