This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00451840_m1
See other SLCO1C1 GE Assays ›
Gene Symbol:
Slco1c1
Gene Name:
solute carrier organic anion transporter family, member 1c1
Gene Aliases:
OATP-14, OATP-F, Oatp2, Oatpf, Slc21a14
Chromosome Location:
Chr.6: 141524378 - 141570177 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00451840_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slco1c1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001177772.1 NP_001171243.1 5 - 6 915 82
NM_021471.2 NP_067446.1 5 - 6 915 82
GenBank mRNA AK045782.1 - 918 82
AK048926.1 - 5 - 6 885 82
AK161010.1 - 885 82
AY007379.1 - 5 - 6 751 82
BC078456.1 - 5 - 6 865 30360663 82

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slco1c1

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
bile acid and bile salt transport
anion transmembrane transport
organic acid transmembrane transport
transporter activity
bile acid transmembrane transporter activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0