This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00452735_m1
See other MSRA GE Assays ›
Gene Symbol:
Msra
Gene Name:
methionine sulfoxide reductase A
Gene Aliases:
2310045J23Rik, 6530413P12Rik, MSR-A
Chromosome Location:
Chr.14: 64122621 - 64455903 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00452735_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Msra
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001253712.1 NP_001240641.1 1 - 2 331 89
NM_001253715.1 NP_001240644.1 1 - 2 317 89
NM_026322.4 NP_080598.2 6 - 7 730 89
GenBank mRNA AK002356.1 - 1 - 2 206 89
AK009822.1 - 1 - 2 208 89
AK030278.1 - 1 - 2 209 89
AK049714.1 - 1 - 2 332 89
BB652761.1 - 1 - 2 329 89
BC014738.1 - 1 - 2 276 4239349 89
BC089311.1 - 6 - 7 730 6412625 89

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1400002231.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Msra

Panther Classification:

Molecular Function -

reductase

Gene Ontology Categories:

Function(s) Process(es)

response to oxidative stress
aging
protein repair
oxidation-reduction process
peptide-methionine (S)-S-oxide reductase activity
oxidoreductase activity
oxidoreductase activity, acting on a sulfur group of donors, disulfide as acceptor

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400017287.mm.1 NM_001253712.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400017287.mm.1 NM_001253712.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400017287.mm.1 NM_001253714.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400017287.mm.1 NM_001253714.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400017287.mm.1 NM_001253715.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400017287.mm.1 NM_001253715.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400017287.mm.1 NM_026322.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400017287.mm.1 NM_026322.4 Refseq
Affymetrix Mouse Clariom S Array PSR1400017287.mm.1 TC1400002231.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400017288.mm.1 NM_001253712.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400017288.mm.1 NM_001253712.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400017288.mm.1 NM_001253714.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400017288.mm.1 NM_001253714.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400017288.mm.1 NM_001253715.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400017288.mm.1 NM_001253715.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400017288.mm.1 NM_026322.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400017288.mm.1 NM_026322.4 Refseq
Affymetrix Mouse Clariom S Array PSR1400017288.mm.1 TC1400002231.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400017291.mm.1 NM_026322.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400017291.mm.1 NM_026322.4 Refseq
Affymetrix Mouse Clariom S Array PSR1400017291.mm.1 TC1400002231.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400017292.mm.1 NM_026322.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400017292.mm.1 NM_026322.4 Refseq
Affymetrix Mouse Clariom S Array PSR1400017292.mm.1 TC1400002231.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400017293.mm.1 NM_026322.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400017293.mm.1 NM_026322.4 Refseq
Affymetrix Mouse Clariom S Array PSR1400017293.mm.1 TC1400002231.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400017290.mm.1 NM_026322.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400017290.mm.1 NM_026322.4 Refseq
Affymetrix Mouse Clariom S Array PSR1400017290.mm.1 TC1400002231.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400017285.mm.1 NM_001253712.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400017285.mm.1 NM_001253712.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400017285.mm.1 NM_001253715.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400017285.mm.1 NM_001253715.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400017285.mm.1 NM_001253716.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400017285.mm.1 NM_001253716.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400017285.mm.1 NM_026322.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400017285.mm.1 NM_026322.4 Refseq
Affymetrix Mouse Clariom S Array PSR1400017285.mm.1 TC1400002231.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 805251 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline