This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00452757_m1
See other MRPS11 GE Assays ›
Gene Symbol:
Mrps11
Gene Name:
mitochondrial ribosomal protein S11
Gene Aliases:
0710005I03Rik, C79873
Chromosome Location:
Chr.7: 78783131 - 78792989 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00452757_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mrps11
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026498.2 NP_080774.2 3 - 4 503 69
GenBank mRNA AB049945.1 - 3 - 4 227 69
AK002993.1 - 3 - 4 340 69
AK152027.1 - 3 - 4 294 69
AK160207.1 - 3 - 4 283 69
AK166241.1 - 3 - 4 291 69
BC005701.1 - 3 - 4 245 69
BC028279.1 - 3 - 4 295 5370270 69

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700001110.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mrps11

Panther Classification:

Molecular Function -

ribosomal protein

Gene Ontology Categories:

Function(s) Process(es)

translation
biological_process
structural constituent of ribosome
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700009751.mm.1 NM_026498.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700009751.mm.1 NM_026498.2 Refseq
Affymetrix Mouse Clariom S Array PSR0700009751.mm.1 TC0700001110.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-wt24k:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline