This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00452895_m1
See other SMUG1 GE Assays ›
Gene Symbol:
Smug1
Gene Name:
single-strand selective monofunctional uracil DNA glycosylase
Gene Aliases:
1200013B09Rik, A930006H09Rik, C85220
Chromosome Location:
Chr.15: 103153290 - 103168895 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00452895_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Smug1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK004735.1 - 1 - 2 65 101
AK172510.1 - 1 - 2 62 101
BC050253.1 - 5 - 6 605 6467286 101
BC054071.1 - 5 - 6 605 101

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500002310.mm.1

Additional Information:

This assay is in the 5'UTR region of the AK004735.1,AK172510.1,BC050253.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

Panther Classification:

Molecular Function -

DNA metabolism protein DNA glycosylase nucleic acid metabolism protein

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500019153.mm.1 TC1500002310.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500019150.mm.1 TC1500002310.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500019155.mm.1 TC1500002310.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500019152.mm.1 TC1500002310.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500019154.mm.1 TC1500002310.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500019141.mm.1 NM_027885.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500019141.mm.1 NM_027885.3 Refseq
Affymetrix Mouse Clariom S Array PSR1500019141.mm.1 TC1500002310.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 890983 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-tks2p:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0