This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00452896_g1
See other SMUG1 GE Assays ›
Gene Symbol:
Smug1
Gene Name:
single-strand selective monofunctional uracil DNA glycosylase
Gene Aliases:
1200013B09Rik, A930006H09Rik, C85220
Chromosome Location:
Chr.15: 103153290 - 103168895 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00452896_g1
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Smug1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_027885.3 NP_082161.2 3 - 4 395 103
GenBank mRNA AK004735.1 - 2 - 3 131 103
AK172510.1 - 2 - 3 128 103
BC050253.1 - 6 - 7 671 6467286 103
BC054071.1 - 6 - 7 671 103
BC062960.1 - 3 - 4 395 30612187 103

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500002310.mm.1

Additional Information:

This assay is in the 5'UTR region of the AK004735.1,AK172510.1,BC050253.1,BC062960.1,NM_027885.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Smug1

Panther Classification:

Molecular Function -

DNA metabolism protein DNA glycosylase nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

DNA repair
base-excision repair
cellular response to DNA damage stimulus
metabolic process
nucleic acid phosphodiester bond hydrolysis
purine-specific mismatch base pair DNA N-glycosylase activity
oxidized base lesion DNA N-glycosylase activity
oxidized pyrimidine nucleobase lesion DNA N-glycosylase activity
DNA binding
single-stranded DNA binding
uracil DNA N-glycosylase activity
hydrolase activity
hydrolase activity, acting on glycosyl bonds
single-strand selective uracil DNA N-glycosylase activity
DNA N-glycosylase activity
DNA-3-methylbase glycosylase activity
nicotinamide riboside hydrolase activity
nicotinic acid riboside hydrolase activity
deoxyribonucleoside 5'-monophosphate N-glycosidase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500019143.mm.1 NM_027885.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500019143.mm.1 NM_027885.3 Refseq
Affymetrix Mouse Clariom S Array PSR1500019143.mm.1 TC1500002310.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500019138.mm.1 NM_027885.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500019138.mm.1 NM_027885.3 Refseq
Affymetrix Mouse Clariom S Array PSR1500019138.mm.1 TC1500002310.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500019145.mm.1 NM_027885.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500019145.mm.1 NM_027885.3 Refseq
Affymetrix Mouse Clariom S Array PSR1500019145.mm.1 TC1500002310.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0