This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00452910_m1
See other NEIL1 GE Assays ›
Gene Symbol:
Neil1
Gene Name:
nei endonuclease VIII-like 1 (E. coli)
Gene Aliases:
2810450N13Rik, Nei1
Chromosome Location:
Chr.9: 57142788 - 57148675 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00452910_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Neil1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_028347.2 NP_082623.1 1 - 2 432 64
GenBank mRNA AB079069.1 - 1 - 2 432 64
AK013322.1 - 2 - 3 503 64
AK155405.1 - 3 - 4 308 64
BC034873.1 - 2 - 3 677 64
BC043297.1 - 2 - 3 596 5320651 64

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900002365.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Neil1

Panther Classification:

Molecular Function -

DNA glycosylase

Gene Ontology Categories:

Function(s) Process(es)

DNA repair
base-excision repair
nucleotide-excision repair
cellular response to DNA damage stimulus
response to oxidative stress
metabolic process
negative regulation of nuclease activity
purine-specific mismatch base pair DNA N-glycosylase activity
nucleic acid binding
DNA binding
damaged DNA binding
catalytic activity
DNA-(apurinic or apyrimidinic site) lyase activity
uracil DNA N-glycosylase activity
protein C-terminus binding
zinc ion binding
hydrolase activity
hydrolase activity, acting on glycosyl bonds
hydrolase activity, hydrolyzing N-glycosyl compounds
lyase activity
single-strand selective uracil DNA N-glycosylase activity
DNA N-glycosylase activity
DNA-3-methylbase glycosylase activity
nicotinamide riboside hydrolase activity
nicotinic acid riboside hydrolase activity
deoxyribonucleoside 5'-monophosphate N-glycosidase activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900019799.mm.1 NM_028347.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900019799.mm.1 NM_028347.2 Refseq
Affymetrix Mouse Clariom S Array PSR0900019799.mm.1 TC0900002365.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900019801.mm.1 NM_028347.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900019801.mm.1 NM_028347.2 Refseq
Affymetrix Mouse Clariom S Array PSR0900019801.mm.1 TC0900002365.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 385524 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline