Assay Details
| Gene Symbol: Neil1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028347.2 | NP_082623.1 | 2 - 3 | 552 | 103 | |
| GenBank mRNA | AB079069.1 | - | 2 - 3 | 552 | 103 | |
| AK013322.1 | - | 3 - 4 | 623 | 103 | ||
| AK155405.1 | - | 4 - 5 | 428 | 103 | ||
| AK186023.1 | - | 1 - 2 | 112 | 103 | ||
| BC034873.1 | - | 3 - 4 | 797 | 103 | ||
| BC043297.1 | - | 3 - 4 | 716 | 5320651 | 103 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
DNA repair
base-excision repair
nucleotide-excision repair
cellular response to DNA damage stimulus
response to oxidative stress
metabolic process
negative regulation of nuclease activity
purine-specific mismatch base pair DNA N-glycosylase activity
nucleic acid binding
DNA binding
damaged DNA binding
catalytic activity
DNA-(apurinic or apyrimidinic site) lyase activity
uracil DNA N-glycosylase activity
protein C-terminus binding
zinc ion binding
hydrolase activity
hydrolase activity, acting on glycosyl bonds
hydrolase activity, hydrolyzing N-glycosyl compounds
lyase activity
single-strand selective uracil DNA N-glycosylase activity
DNA N-glycosylase activity
DNA-3-methylbase glycosylase activity
nicotinamide riboside hydrolase activity
nicotinic acid riboside hydrolase activity
deoxyribonucleoside 5'-monophosphate N-glycosidase activity
DNA repair
base-excision repair
nucleotide-excision repair
cellular response to DNA damage stimulus
response to oxidative stress
metabolic process
negative regulation of nuclease activity
purine-specific mismatch base pair DNA N-glycosylase activity
nucleic acid binding
DNA binding
damaged DNA binding
catalytic activity
DNA-(apurinic or apyrimidinic site) lyase activity
uracil DNA N-glycosylase activity
protein C-terminus binding
zinc ion binding
hydrolase activity
hydrolase activity, acting on glycosyl bonds
hydrolase activity, hydrolyzing N-glycosyl compounds
lyase activity
single-strand selective uracil DNA N-glycosylase activity
DNA N-glycosylase activity
DNA-3-methylbase glycosylase activity
nicotinamide riboside hydrolase activity
nicotinic acid riboside hydrolase activity
deoxyribonucleoside 5'-monophosphate N-glycosidase activity
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2019615 | BC043297.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2019615 | AK155405.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2019615 | AK013322.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2099415 | BC043297.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2099415 | AK155405.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2099415 | BC034873.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2099415 | NM_028347.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2099415 | AB079069.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 19852 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 757312 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P111010 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2099415 | BC043297.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2099415 | AK155405.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2099415 | BC034873.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2099415 | NM_028347.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2099415 | AB079069.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P109741 | BC043297.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P109741 | AK155405.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P109741 | BC034873.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P109741 | AK013322.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P109742 | AK013322.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26540545 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26540570 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26540558 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26540538 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26540578 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26540565 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26540542 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26540551 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26540526 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26540550 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26540559 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26540576 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26540547 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26540531 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26540549 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26540529 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26540567 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26540563 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26540541 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26540557 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26540566 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26540533 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26540535 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26540575 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26540553 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26540527 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26540581 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26540568 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26540554 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26540540 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26540572 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P111010 | - | - |
| Whole Mouse Genome G4122A | A_51_P109742 | AK013322.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P109741 | BC043297.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P109741 | AK155405.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P109741 | BC034873.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P109741 | AK013322.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P106433 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P114905 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P111010 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07514837 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07514870 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07514858 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07958009 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07514848 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07514857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26540523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26540568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26540551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26540444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26540459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26540588 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26540442 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26540466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26540566 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26540437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26540468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26540557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26540518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26540586 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26540554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26540448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26540519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26540583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26540578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26540538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26540559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26540439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26540455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26540526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26540469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26540533 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26540572 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26540524 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26540599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26540564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26540458 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26540553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26540563 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26540516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26540597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26540464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26540513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26540549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26540445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26540589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26540540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26540542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26540545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26540456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26540451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26540470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26540461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26540593 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26540462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26540520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26540529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26540590 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26540443 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26540535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26540547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26540584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26540527 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26540600 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26540457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26540558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26540454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26540567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26540575 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07612934 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26540531 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26540547 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26540572 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26540582 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07988260 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26540552 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07899295 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26540551 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26540511 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26540557 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26540575 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26540576 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26540559 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26540558 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26540520 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26540527 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26540570 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26540571 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07514859 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26540519 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26540524 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26540549 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26540526 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07988274 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26540533 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07988264 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26540554 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07514868 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26540523 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07514864 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26540567 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26540573 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26540518 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26540568 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26540542 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07883060 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26540516 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26540541 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07923548 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26540530 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26540545 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26540540 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07580741 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10593890 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10593898 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10593888 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10593902 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10593897 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10593889 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10593892 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10593900 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10593901 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10593893 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10593895 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10593894 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10593896 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10593899 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10593891 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1452871_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10593887 | - | - |
| Mouse Genome 430 2.0 Array | 1452871_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1452871_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900019798.mm.1 | NM_028347.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900019798.mm.1 | NM_028347.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900019798.mm.1 | TC0900002365.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1452871_PM_at | - | - |
| Mouse Expression Set 430 | 1452871_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1452871_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1452871_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 105670458 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 105670458 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2640341 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002640341 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2632630 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2632630 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2561392 | - | - |
| mouse-6 v1.1 (Illumina) | scl35798.7.1_9-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 105670458 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2640341 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl35798.7.1_9-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2640341 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 105670458 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2632630 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2561392 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2632630 | - | - |

Neil1
Genomic Map