This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00452923_m1
See other NPL GE Assays ›
Gene Symbol:
Npl
Gene Name:
N-acetylneuraminate pyruvate lyase
Gene Aliases:
0610033B02Rik
Chromosome Location:
Chr.1: 153503016 - 153549714 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00452923_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Npl
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_028749.1 NP_083025.1 4 - 5 316 65
GenBank mRNA AK002734.1 - 4 - 5 316 65
AK088859.1 - 4 - 5 318 65
BC022734.1 - 4 - 5 312 4506776 65

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100003280.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Npl

Panther Classification:

Molecular Function -

lyase

Gene Ontology Categories:

Function(s) Process(es)

carbohydrate metabolic process
metabolic process
N-acetylneuraminate catabolic process
catalytic activity
N-acetylneuraminate lyase activity
lyase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100026698.mm.1 NM_028749.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100026698.mm.1 NM_028749.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100026698.mm.1 TC0100003280.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0