This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00453704_m1
See other TMPRSS3 GE Assays ›
Gene Symbol:
Tmprss3
Gene Name:
transmembrane protease, serine 3
Gene Aliases:
-
Chromosome Location:
Chr.17: 31179265 - 31209743 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00453704_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tmprss3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001163776.1 NP_001157248.1 12 - 13 1542 68
NM_080727.2 NP_542765.2 11 - 12 1402 68
GenBank mRNA AJ300738.1 - 11 - 12 1402 68
AK158203.1 - 12 - 13 1545 68
BC112375.1 - 11 - 12 1399 40057032 68

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1700001814.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Protease 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tmprss3

Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
cellular sodium ion homeostasis
sensory perception of sound
serine-type endopeptidase activity
scavenger receptor activity
peptidase activity
serine-type peptidase activity
hydrolase activity
sodium channel regulator activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2011151 AK158203.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2011151 NM_080727.2 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2011151 NM_001163776.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2138600 BC112375.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2138600 AJ300738.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2138600 AK158203.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2138600 NM_080727.2 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2138600 NM_001163776.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 123517 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 124027 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 21756 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 910302 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 5128 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 24109 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 307653 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 38699 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P154391 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P175994 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2011151 AK158203.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2011151 NM_080727.2 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2011151 NM_001163776.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2138600 BC112375.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2138600 AJ300738.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2138600 AK158203.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2138600 NM_080727.2 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2138600 NM_001163776.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P362298 AJ300738.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P362298 AK158203.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P362298 NM_080727.2 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P362298 NM_001163776.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31138364 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31138371 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31138382 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31138370 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31138386 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31138374 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31138383 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31138368 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31138375 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31138367 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31138377 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31138373 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31138379 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P175994 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P154391 - -
Whole Mouse Genome G4122A A_51_P362298 AJ300738.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P362298 AK158203.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P362298 NM_080727.2 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P362298 NM_001163776.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P123817 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P118874 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P175994 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P154391 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13086403 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13086395 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31138379 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31138382 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31138373 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31138370 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31138375 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31138377 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31138374 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10449681 - -
Affymetrix Mouse Gene 1.0 ST Array 10449673 - -
Affymetrix Mouse Gene 1.0 ST Array 10449677 - -
Affymetrix Mouse Gene 1.0 ST Array 10449679 - -
Affymetrix Mouse Gene 1.0 ST Array 10449676 - -
Affymetrix Mouse Gene 1.0 ST Array 10449678 - -
Affymetrix Mouse Gene 1.0 ST Array 10449682 - -
Affymetrix Mouse Gene 1.0 ST Array 10449684 - -
Affymetrix Mouse Gene 1.0 ST Array 10449685 - -
Affymetrix Mouse Gene 1.0 ST Array 10449683 - -
Affymetrix Mouse Gene 1.0 ST Array 10449680 - -
Affymetrix Mouse Gene 1.0 ST Array 10449674 - -
Affymetrix Mouse Gene 1.0 ST Array 10449675 - -
Affymetrix HT MG-430 PM Array Plate 1421714_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10449672 - -
Mouse Genome 430 2.0 Array 1421714_at - -
Mouse Genome 430 2.0 Array 1421714_PM_at - -
Affymetrix Mouse Clariom S Array PSR1700016674.mm.1 NM_001163776.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700016674.mm.1 NM_001163776.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700016674.mm.1 NM_080727.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700016674.mm.1 NM_080727.2 Refseq
Affymetrix Mouse Clariom S Array PSR1700016674.mm.1 TC1700001814.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1700016673.mm.1 NM_001163776.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700016673.mm.1 NM_001163776.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700016673.mm.1 NM_080727.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700016673.mm.1 NM_080727.2 Refseq
Affymetrix Mouse Clariom S Array PSR1700016673.mm.1 TC1700001814.mm.1 MTA TC
Mouse Expression Set 430 1421714_PM_at - -
Mouse Expression Set 430 1421714_at - -
Mouse Genome 430A 2.0 Array 1421714_PM_at - -
Mouse Genome 430A 2.0 Array 1421714_at - -
Affymetrix HT Mouse Genome 430A Array 1421714_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1421714_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 849986 - -
Array Name Probe IDs Targets Target Type
Illumina mouse-6 v1.0 expression beadchip 6370433 - -
Illumina mouseRef-8 v1.0 expression beadchip 0006370433 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_1243162 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1243162 - -
mouse-6 v1.1 (Illumina) scl50082.7.1_105-S - -
Illumina Sentrix mouse-6.v1 bead array 6370433 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl50082.7.1_105-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 6370433 - -
MouseRef-8 v2 Expression BeadChip ILMN_1243162 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_1243162 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0