This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00455403_g1
See other PRSS50 GE Assays ›
Gene Symbol:
Prss50
Gene Name:
protease, serine 50
Gene Aliases:
79H19C, Tsp50
Chromosome Location:
Chr.9: 110857899 - 110864629 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00455403_g1
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Prss50
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_146227.4 NP_666339.2 4 - 5 1100 84
GenBank mRNA AF481144.1 - 4 - 5 1086 84
AK045168.1 - 4 - 5 1101 84
BC150686.1 - 4 - 5 1099 9087597 84

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900001474.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Prss50

Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
serine-type endopeptidase activity
threonine-type endopeptidase activity
peptidase activity
hydrolase activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26826271 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26826274 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26826273 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26826270 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26826272 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2079787 AK045168.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2079787 BC150686.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2079787 NM_146227.4 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2079787 AF481144.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2176748 AK045168.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2176748 BC150686.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2176748 NM_146227.4 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2176748 AF481144.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 792862 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 840925 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 290404 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 82756 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P105599 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2176748 AK045168.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2176748 BC150686.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2176748 NM_146227.4 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2176748 AF481144.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2079787 AK045168.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2079787 BC150686.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2079787 NM_146227.4 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2079787 AF481144.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26826264 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26826274 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26826267 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26826280 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26826263 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26826261 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26826270 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26826277 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26826276 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26826271 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26826282 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26826273 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26826268 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26826278 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26826266 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P105599 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02029763 - -
Whole Mouse Genome G4122A A_51_P339803 AK045168.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P339803 BC150686.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P339803 NM_146227.4 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P339803 AF481144.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P678096 AK045168.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P678096 BC150686.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P678096 NM_146227.4 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P678096 AF481144.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134354 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P105599 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08139281 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07794263 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07794255 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08139282 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07794257 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07471078 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26826272 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26826271 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26826273 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26826270 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26826274 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826392 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826367 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826380 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826277 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826391 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826300 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826308 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826385 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826364 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826298 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826296 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826263 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826261 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826305 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826284 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826224 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826377 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826309 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826233 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826271 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826384 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826226 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826374 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826289 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826267 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826229 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826270 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826290 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826274 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826310 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826264 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826381 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826397 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826376 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826362 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826365 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826315 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826282 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826386 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826382 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826318 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826312 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826389 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826273 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826230 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826369 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826394 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826387 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826276 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826372 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826393 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826291 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826278 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826294 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826302 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826399 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826314 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26826285 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26826288 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26826318 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08139282 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26826314 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07794260 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26826291 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26826276 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07876132 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07794254 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07964091 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26826290 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26826310 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07471078 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26826286 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07964105 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26826266 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26826315 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26826278 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26826312 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26826304 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07876129 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07876127 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26826289 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26826270 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26826309 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07964111 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26826305 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07964110 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26826296 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26826282 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26826279 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07471082 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07964103 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10589651 - -
Affymetrix Mouse Gene 1.0 ST Array 10589653 - -
Affymetrix Mouse Gene 1.0 ST Array 10589649 - -
Affymetrix Mouse Gene 1.0 ST Array 10589652 - -
Affymetrix Mouse Gene 1.0 ST Array 10589650 - -
Affymetrix Mouse Gene 1.0 ST Array 10589648 - -
Affymetrix Mouse Gene 1.1 ST Array 10589647 - -
Mouse Genome 430 2.0 Array 1439739_PM_at - -
Mouse Genome 430 2.0 Array 1439739_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 235631_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 235631_at - -
Affymetrix Mouse Clariom S Array PSR0900012338.mm.1 NM_146227.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900012338.mm.1 NM_146227.4 Refseq
Affymetrix Mouse Clariom S Array PSR0900012338.mm.1 TC0900001474.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900012339.mm.1 NM_146227.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900012339.mm.1 NM_146227.4 Refseq
Affymetrix Mouse Clariom S Array PSR0900012339.mm.1 TC0900001474.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900012341.mm.1 NM_146227.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900012341.mm.1 NM_146227.4 Refseq
Affymetrix Mouse Clariom S Array PSR0900012341.mm.1 TC0900001474.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900012337.mm.1 NM_146227.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900012337.mm.1 NM_146227.4 Refseq
Affymetrix Mouse Clariom S Array PSR0900012337.mm.1 TC0900001474.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900012342.mm.1 NM_146227.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900012342.mm.1 NM_146227.4 Refseq
Affymetrix Mouse Clariom S Array PSR0900012342.mm.1 TC0900001474.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900012340.mm.1 NM_146227.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900012340.mm.1 NM_146227.4 Refseq
Affymetrix Mouse Clariom S Array PSR0900012340.mm.1 TC0900001474.mm.1 MTA TC
Mouse Expression Set 430 1439739_PM_at - -
Mouse Expression Set 430 1439739_at - -
Murine Genome U74v2 Set 168974_r_at - -
Murine Genome U74v2 Set 168974_PM_r_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 357269 - -
Array Name Probe IDs Targets Target Type
Illumina mouse-6 v1.0 expression beadchip 6100593 - -
Illumina mouseRef-8 v1.0 expression beadchip 0006100593 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_1245792 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1245792 - -
mouse-6 v1.1 (Illumina) scl36417.5.1_79-S - -
Illumina Sentrix mouse-6.v1 bead array 6100593 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl36417.5.1_79-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 6100593 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_1245792 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0