This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00455757_m1
See other SLCO4A1 GE Assays ›
Gene Symbol:
Slco4a1
Gene Name:
solute carrier organic anion transporter family, member 4a1
Gene Aliases:
OATP-E, Slc21a12
Chromosome Location:
Chr.2: 180456250 - 180474853 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00455757_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slco4a1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_148933.1 NP_683735.1 5 - 6 1434 79
GenBank mRNA AK033598.1 - 5 - 6 1254 79
AK172688.1 - 6 - 7 1357 79
BC030719.1 - 5 - 6 1434 4501966 79
BC030720.1 - 5 - 6 1383 4505508 79

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200002792.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slco4a1

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
organic anion transport
thyroid hormone metabolic process
thyroid hormone transport
anion transmembrane transport
transporter activity
organic anion transmembrane transporter activity
thyroid hormone transmembrane transporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200022827.mm.1 NM_148933.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200022827.mm.1 NM_148933.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200022827.mm.1 TC0200002792.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200022829.mm.1 NM_148933.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200022829.mm.1 NM_148933.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200022829.mm.1 TC0200002792.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0