This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00456361_m1
See other TPO GE Assays ›
Gene Symbol:
Tpo
Gene Name:
thyroid peroxidase
Gene Aliases:
-
Chromosome Location:
Chr.12: 30054661 - 30132984 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00456361_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tpo
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009417.2 NP_033443.1 14 - 15 2556 73
GenBank mRNA AK047843.1 - 14 - 15 2535 73
X60703.1 - 14 - 15 2556 73

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1200001580.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tpo

Panther Classification:

Molecular Function -

peroxidase

Gene Ontology Categories:

Function(s) Process(es)

thyroid hormone generation
response to oxidative stress
embryonic hemopoiesis
hormone biosynthetic process
hydrogen peroxide catabolic process
oxidation-reduction process
iodide peroxidase activity
peroxidase activity
calcium ion binding
oxidoreductase activity
heme binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1200010934.mm.1 NM_009417.3 Refseq
Affymetrix Mouse Clariom S Array PSR1200010934.mm.1 TC1200001580.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1200010933.mm.1 NM_009417.3 Refseq
Affymetrix Mouse Clariom S Array PSR1200010933.mm.1 TC1200001580.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0