This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00456720_m1
See other HAL GE Assays ›
Gene Symbol:
Hal
Gene Name:
histidine ammonia lyase
Gene Aliases:
Hsd, his, histidase
Chromosome Location:
Chr.10: 93488768 - 93516743 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00456720_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Hal
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010401.3 NP_034531.1 19 - 20 2198 104
GenBank mRNA AK014518.1 - 19 - 20 2198 104
AK017397.1 - 4 - 5 372 104
AK028963.1 - 19 - 20 1905 104
AK081409.1 - 19 - 20 1901 104
BC057637.1 - 19 - 20 1875 4211545 104
BC157999.1 - 19 - 20 2180 9088147 104
L07645.1 - 19 - 20 1882 104

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000001133.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Hal

Panther Classification:

Molecular Function -

lyase

Gene Ontology Categories:

Function(s) Process(es)

histidine metabolic process
histidine catabolic process
biosynthetic process
catalytic activity
histidine ammonia-lyase activity
lyase activity
ammonia-lyase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000008575.mm.1 NM_010401.4 Refseq
Affymetrix Mouse Clariom S Array PSR1000008575.mm.1 TC1000001133.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0