This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00456921_m1
See other SNAP25 GE Assays ›
Gene Symbol:
Snap25
Gene Name:
synaptosomal-associated protein 25
Gene Aliases:
Bdr, GENA70, SNAP-25, SUP, sp
Chromosome Location:
Chr.2: 136713438 - 136782428 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00456921_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Snap25
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011428.3 NP_035558.1 4 - 5 376 71
GenBank mRNA AF483516.1 - 3 - 4 163 71
AF483517.1 - 3 - 4 163 71
AK078038.1 - 4 - 5 374 71
BC018249.1 - 4 - 5 264 4504644 71
M22012.1 - 4 - 5 326 71

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200002077.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Snap25

Panther Classification:

Molecular Function -

SNARE protein

Gene Ontology Categories:

Function(s) Process(es)

exocytosis
neurotransmitter secretion
long-term memory
locomotory behavior
associative learning
regulation of neuron projection development
synaptic vesicle priming
endosomal transport
sleep
synaptic vesicle fusion to presynaptic active zone membrane
regulation of ion transmembrane transport
positive regulation of hormone secretion
calcium ion-regulated exocytosis of neurotransmitter
long-term synaptic potentiation
regulation of establishment of protein localization
potassium ion transmembrane transport
SNARE binding
voltage-gated potassium channel activity
SNAP receptor activity
protein binding
myosin binding
syntaxin-1 binding
protein domain specific binding
ion channel binding
protein N-terminus binding
calcium-dependent protein binding

MicroArray Probes:

ABI

Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 535596 - -
ABI Mouse Genome Survey Microarray 597507 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-55658555d6-h6m8v:80/100.66.79.224:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline