This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00457083_g1
See other VRK1 GE Assays ›
Gene Symbol:
Vrk1
Gene Name:
vaccinia related kinase 1
Gene Aliases:
51PK
Chromosome Location:
Chr.12: 106010163 - 106077421 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00457083_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Vrk1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001029843.1 NP_001025014.1 2 - 3 264 60
NM_001029844.1 NP_001025015.1 2 - 3 264 60
NM_011705.3 NP_035835.1 2 - 3 264 60
GenBank mRNA AF080252.1 - 1 - 2 177 60
AF080253.1 - 2 - 3 218 60
AK032344.1 - 2 - 3 235 60
AK088230.1 - 2 - 3 194 60
AK088674.1 - 2 - 3 247 60
AK136352.1 - 2 - 3 238 60
AK166676.1 - 2 - 3 232 60
BC016676.1 - 2 - 3 299 3602292 60
BC051072.1 - 2 - 3 244 60
BY130767.1 - 2 - 3 264 60

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1200001077.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Vrk1

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
endocytosis
cell cycle
mitotic nuclear division
regulation of cell shape
Wnt signaling pathway
phosphorylation
peptidyl-serine phosphorylation
histone H3-S10 phosphorylation
protein autophosphorylation
cell division
histone H3-T3 phosphorylation
Golgi disassembly
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
protein kinase binding
nucleosomal histone binding
histone kinase activity (H3-S10 specific)
histone kinase activity (H3-T3 specific)

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1200007739.mm.1 NM_001029843.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200007739.mm.1 NM_001029843.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200007739.mm.1 NM_001029844.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200007739.mm.1 NM_001029844.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200007739.mm.1 NM_011705.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200007739.mm.1 NM_011705.3 Refseq
Affymetrix Mouse Clariom S Array PSR1200007739.mm.1 TC1200001077.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline