This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00457580_m1
See other ADGRG1 GE Assays ›
Gene Symbol:
Adgrg1
Gene Name:
adhesion G protein-coupled receptor G1
Gene Aliases:
Cyt28, Gpr56, TM7LN4, TM7XN1
Chromosome Location:
Chr.8: 94974756 - 95014208 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00457580_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Adgrg1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001198894.1 NP_001185823.1 2 - 3 389 88
NM_018882.3 NP_061370.2 2 - 3 339 88
GenBank mRNA AF166382.1 - 2 - 3 389 88
AK085664.1 - 2 - 3 193 88
AK087268.1 - 2 - 3 141 88
AK133678.1 - 2 - 3 106 88
AK141886.1 - 2 - 3 368 88
AK141894.1 - 2 - 3 152 88
AK164776.1 - 2 - 3 163 88
AK167547.1 - 1 - 2 93 88
BC034678.1 - 2 - 3 135 3709247 88

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800001126.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Adgrg1

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

angiogenesis
cell adhesion
signal transduction
cell surface receptor signaling pathway
G-protein coupled receptor signaling pathway
Rho protein signal transduction
multicellular organism development
nervous system development
brain development
negative regulation of cell proliferation
vascular endothelial growth factor production
cerebral cortex regionalization
cerebral cortex radial glia guided migration
layer formation in cerebral cortex
cell differentiation
positive regulation of Rho protein signal transduction
positive regulation of cell adhesion
protein kinase C signaling
positive regulation of neural precursor cell proliferation
negative regulation of neuron migration
signal transducer activity
transmembrane signaling receptor activity
G-protein coupled receptor activity
collagen binding
extracellular matrix binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800008806.mm.1 NM_018882.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800008806.mm.1 NM_018882.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800008806.mm.1 TC0800001126.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800008807.mm.1 NM_018882.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800008807.mm.1 NM_018882.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800008807.mm.1 TC0800001126.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800008810.mm.1 NM_001198894.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800008810.mm.1 NM_001198894.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800008810.mm.1 NM_018882.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800008810.mm.1 NM_018882.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800008810.mm.1 TC0800001126.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 528616 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0