This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00457586_g1
See other ADGRG1 GE Assays ›
Gene Symbol:
Adgrg1
Gene Name:
adhesion G protein-coupled receptor G1
Gene Aliases:
Cyt28, Gpr56, TM7LN4, TM7XN1
Chromosome Location:
Chr.8: 94974756 - 95014208 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00457586_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Adgrg1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001198894.1 NP_001185823.1 8 - 9 1382 86
NM_018882.3 NP_061370.2 8 - 9 1332 86
GenBank mRNA AF166382.1 - 8 - 9 1382 86
AK087268.1 - 8 - 9 1134 86
AK133678.1 - 8 - 9 1099 86
AK141886.1 - 8 - 9 1361 86
AK141894.1 - 8 - 9 1145 86
AK164776.1 - 8 - 9 1157 86
AK167547.1 - 7 - 8 1086 86
AK178910.1 - 1 - 2 40 86
BC034678.1 - 8 - 9 1128 3709247 86

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800001126.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Adgrg1

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

angiogenesis
cell adhesion
signal transduction
cell surface receptor signaling pathway
G-protein coupled receptor signaling pathway
Rho protein signal transduction
multicellular organism development
nervous system development
brain development
negative regulation of cell proliferation
vascular endothelial growth factor production
cerebral cortex regionalization
cerebral cortex radial glia guided migration
layer formation in cerebral cortex
cell differentiation
positive regulation of Rho protein signal transduction
positive regulation of cell adhesion
protein kinase C signaling
positive regulation of neural precursor cell proliferation
negative regulation of neuron migration
signal transducer activity
transmembrane signaling receptor activity
G-protein coupled receptor activity
collagen binding
extracellular matrix binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800008816.mm.1 NM_001198894.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800008816.mm.1 NM_001198894.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800008816.mm.1 NM_018882.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800008816.mm.1 NM_018882.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800008816.mm.1 TC0800001126.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-z4mpm:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0