This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00457633_g1
See other DNMT3L GE Assays ›
Gene Symbol:
Dnmt3l
Gene Name:
DNA (cytosine-5-)-methyltransferase 3-like
Gene Aliases:
D6Ertd14e, ecat7
Chromosome Location:
Chr.10: 78042287 - 78063622 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00457633_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dnmt3l
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001081695.2 NP_001075164.1 11 - 12 1162 63
NM_001284197.1 NP_001271126.1 9 - 10 1003 63
NM_001284198.1 NP_001271127.1 3 - 4 799 63
NM_001284199.1 NP_001271128.1 3 - 4 626 63
NM_001284200.1 NP_001271129.1 2 - 3 460 63
NM_019448.4 NP_062321.1 10 - 11 1016 63
GenBank mRNA AF220524.1 - 9 - 10 947 63
AJ404467.1 - 9 - 10 988 63
AK010434.1 - 10 - 11 1017 63
BC083147.1 - 9 - 10 1001 30447906 63
EF051621.1 - 3 - 4 799 63
EF051622.1 - 3 - 4 626 63
EF051623.1 - 2 - 3 460 63
EF051624.1 - 11 - 12 1162 63

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000000776.mm.1

Additional Information:

This assay is in the 5'UTR region of the NM_001284198.1,NM_001284199.1,NM_001284200.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Dnmt3l

Panther Classification:

Molecular Function -

DNA methyltransferase

Gene Ontology Categories:

Function(s) Process(es)

in utero embryonic development
placenta development
methylation
positive regulation of catalytic activity
negative regulation of transcription, DNA-templated
chorionic trophoblast cell differentiation
genetic imprinting
protein binding
enzyme activator activity
enzyme binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000005691.mm.1 NM_001284198.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000005691.mm.1 NM_001284199.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000005691.mm.1 NM_001284200.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000005691.mm.1 TC1000000776.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1000005695.mm.1 NM_001081695.2 Refseq
Affymetrix Mouse Clariom S Array PSR1000005695.mm.1 NM_001284197.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000005695.mm.1 NM_001284198.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000005695.mm.1 NM_001284199.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000005695.mm.1 NM_001284200.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000005695.mm.1 NM_019448.4 Refseq
Affymetrix Mouse Clariom S Array PSR1000005695.mm.1 TC1000000776.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1000005692.mm.1 NM_001284198.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000005692.mm.1 TC1000000776.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1000005693.mm.1 NM_001284198.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000005693.mm.1 NM_001284199.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000005693.mm.1 TC1000000776.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0